Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00469848_m1
          See other TFPT GE Assays ›
          Gene Symbol:
          Tfpt
          Gene Name:
          TCF3 (E2A) fusion partner
          Gene Aliases:
          2400004F01Rik, AI450389, Amida, FB1
          Chromosome Location:
          Chr.7: 3620324 - 3629929 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00469848_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tfpt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290381.1 NP_001277310.1 2 - 3 637 81
          NM_023524.2 NP_076013.1 2 - 3 637 81
          GenBank mRNA AB035134.1 - 2 - 3 328 81
          AF267988.1 - 2 - 3 592 81
          AK010279.1 - 2 - 3 295 81
          AK155925.1 - 2 - 3 621 81
          BC024356.1 - 2 - 3 536 5066740 81
          CX768547.1 - 1 - 2 156 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002132.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tfpt

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          DNA recombination
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          apoptotic process
          cellular response to DNA damage stimulus
          positive regulation of apoptotic process
          apoptotic signaling pathway
          DNA binding
          protein kinase binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946431 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946432 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946433 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030128 AK010279.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030128 AK155925.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030128 BC024356.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030128 AB035134.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030128 AF267988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030126 AK155925.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030126 BC024356.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030126 AB035134.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030126 AF267988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030126 NM_023524.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444095 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 710221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 433337 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397253 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960124 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367799 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 175534 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030128 AK010279.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030128 AK155925.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030128 BC024356.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030128 AB035134.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030128 AF267988.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030126 AK155925.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030126 BC024356.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030126 AB035134.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030126 AF267988.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030126 NM_023524.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946381 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946385 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946430 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06112379 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06112387 - -
          Whole Mouse Genome G4122A A_52_P601036 AK010279.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P601036 AK155925.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P601036 BC024356.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P601036 AB035134.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P601036 AF267988.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116816 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06112387 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946433 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946432 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946431 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10559410 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559411 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559412 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559413 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559414 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559407 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559409 - -
          Affymetrix HT MG-430 PM Array Plate 1448040_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1418064_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438963_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1438964_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10559406 - -
          Mouse Genome 430 2.0 Array 1438963_PM_s_at - -
          Mouse Genome 430 2.0 Array 1448040_PM_at - -
          Mouse Genome 430 2.0 Array 1438963_s_at - -
          Mouse Genome 430 2.0 Array 1438964_PM_x_at - -
          Mouse Genome 430 2.0 Array 1418064_at - -
          Mouse Genome 430 2.0 Array 1438964_x_at - -
          Mouse Genome 430 2.0 Array 1448040_at - -
          Mouse Genome 430 2.0 Array 1418064_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69714_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69714_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700019192.mm.1 TC0700002132.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019192.mm.1 NM_023524.2 Refseq
          Mouse Expression Set 430 1438963_s_at - -
          Mouse Expression Set 430 1448040_PM_at - -
          Mouse Expression Set 430 1418064_at - -
          Mouse Expression Set 430 1418064_PM_at - -
          Mouse Expression Set 430 1448040_at - -
          Mouse Expression Set 430 1438964_PM_x_at - -
          Mouse Expression Set 430 1438964_x_at - -
          Mouse Expression Set 430 1438963_PM_s_at - -
          Murine Genome U74v2 Set 160928_PM_at - -
          Murine Genome U74v2 Set 160928_at - -
          Mouse Genome 430A 2.0 Array 1448040_at - -
          Mouse Genome 430A 2.0 Array 1438963_s_at - -
          Mouse Genome 430A 2.0 Array 1438964_x_at - -
          Mouse Genome 430A 2.0 Array 1438964_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1438963_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1418064_at - -
          Mouse Genome 430A 2.0 Array 1448040_PM_at - -
          Mouse Genome 430A 2.0 Array 1418064_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418064_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418064_at - -
          Affymetrix HT Mouse Genome 430A Array 1438964_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1448040_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448040_at - -
          Affymetrix HT Mouse Genome 430A Array 1438963_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1438963_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1438964_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 477016 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6450239 - -
          MouseWG-6 v2 Expression BeadChip 5860309 - -
          Illumina mouse-6 v1.0 expression beadchip 5860309 - -
          Illumina mouse-6 v1.0 expression beadchip 6450239 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006450239 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005860309 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1217000 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222401 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222401 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217000 - -
          mouse-6 v1.1 (Illumina) scl31834.6.1_1-S - -
          mouse-6 v1.1 (Illumina) scl000124.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 6450239 - -
          Illumina Sentrix mouse-6.v1 bead array 5860309 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000124.1_0-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31834.6.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6450239 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5860309 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1217000 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2877706 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222401 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1217000 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222401 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2877706 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000124.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl31834.6.1_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline