Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00470020_g1
          See other LIME1 GE Assays ›
          Gene Symbol:
          Lime1
          Gene Name:
          Lck interacting transmembrane adaptor 1
          Gene Aliases:
          2810038K19Rik, LIME
          Chromosome Location:
          Chr.2: 181381235 - 181383628 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00470020_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lime1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023684.2 NP_076173.1 2 - 3 1138 93
          GenBank mRNA AF115339.1 - 2 - 3 1138 93
          AK012881.1 - 2 - 3 202 93
          AK172632.1 - 2 - 3 84 93
          BC023065.1 - 2 - 3 482 5357176 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002822.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK172632.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lime1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          regulation of transcription from RNA polymerase II promoter
          regulation of phosphatidylinositol 3-kinase signaling
          regulation of NF-kappaB import into nucleus
          regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of MAP kinase activity
          T cell receptor signaling pathway
          B cell receptor signaling pathway
          regulation of release of sequestered calcium ion into cytosol
          protein binding
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102419 AF115339.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102419 BC023065.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102419 NM_023684.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974452 AF115339.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974452 AK012881.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974452 AK172632.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974452 BC023065.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974452 NM_023684.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144606 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139788 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128557 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102419 AF115339.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102419 BC023065.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102419 NM_023684.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974452 AF115339.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974452 AK012881.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974452 AK172632.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974452 BC023065.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974452 NM_023684.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21910274 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128557 - -
          Whole Mouse Genome G4122A A_51_P212713 AF115339.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212713 BC023065.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212713 NM_023684.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121049 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128557 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01655587 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01655568 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01655571 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01655575 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01655582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21910302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01655602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02142237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02142222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01655564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01382536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01655581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01655548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01850287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01850288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01850291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01655545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21910320 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10479611 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479608 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479613 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479614 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479612 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479609 - -
          Affymetrix HT MG-430 PM Array Plate 1437589_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1448500_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1416869_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10479607 - -
          Mouse Genome 430 2.0 Array 1437589_x_at - -
          Mouse Genome 430 2.0 Array 1416869_x_at - -
          Mouse Genome 430 2.0 Array 1448500_a_at - -
          Mouse Genome 430 2.0 Array 1437589_PM_x_at - -
          Mouse Genome 430 2.0 Array 1416869_PM_x_at - -
          Mouse Genome 430 2.0 Array 1448500_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72699_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72699_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200023262.mm.1 NM_023684.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023262.mm.1 NM_023684.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023262.mm.1 TC0200002822.mm.1 MTA TC
          Mouse Expression Set 430 1424548_at - -
          Mouse Expression Set 430 1416869_x_at - -
          Mouse Expression Set 430 1437589_x_at - -
          Mouse Expression Set 430 1424548_PM_at - -
          Mouse Expression Set 430 1416869_PM_x_at - -
          Mouse Expression Set 430 1448500_PM_a_at - -
          Mouse Expression Set 430 1448500_a_at - -
          Mouse Expression Set 430 1437589_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1416869_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1448500_a_at - -
          Mouse Genome 430A 2.0 Array 1448500_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1424548_at - -
          Mouse Genome 430A 2.0 Array 1424548_PM_at - -
          Mouse Genome 430A 2.0 Array 1416869_x_at - -
          Mouse Genome 430A 2.0 Array 1437589_x_at - -
          Mouse Genome 430A 2.0 Array 1437589_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 467017 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3850403 - -
          Illumina mouse-6 v1.0 expression beadchip 3850403 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003850403 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238893 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238893 - -
          mouse-6 v1.1 (Illumina) scl072699.4_22-S - -
          Illumina Sentrix mouse-6.v1 bead array 3850403 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl072699.4_22-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3850403 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1238893 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238893 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl072699.4_22-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline