Search
Search
| Gene Symbol: Ppbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023785.2 | NP_076274.1 | 1 - 2 | 209 | 62 | |
| GenBank mRNA | AB042817.1 | - | 1 - 2 | 218 | 62 | |
| AF219112.1 | - | 1 - 2 | 209 | 62 | ||
| AF278700.1 | - | 1 - 2 | 209 | 62 | ||
| AK010273.1 | - | 1 - 2 | 210 | 62 | ||
| AK157077.1 | - | 1 - 2 | 209 | 62 | ||
| AK157141.1 | - | 1 - 2 | 210 | 62 | ||
| AK158061.1 | - | 1 - 2 | 207 | 62 | ||
| AK171720.1 | - | 1 - 2 | 210 | 62 | ||
| BC127043.1 | - | 1 - 2 | 176 | 40053104 | 62 | |
| BC127044.1 | - | 1 - 2 | 176 | 40053105 | 62 | |
Set Membership: |
|