Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00470322_m1
          See other GDPD3 GE Assays ›
          Gene Symbol:
          Gdpd3
          Gene Name:
          glycerophosphodiester phosphodiesterase domain containing 3
          Gene Aliases:
          1110015E22Rik
          Chromosome Location:
          Chr.7: 126766414 - 126775645 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00470322_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gdpd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024228.2 NP_077190.2 7 - 8 765 61
          GenBank mRNA AK003726.1 - 7 - 8 767 61
          AW045753.1 - 3 - 4 61
          BC002172.1 - 5 - 6 684 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001815.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gdpd3

          Panther Classification:

          Molecular Function -

          phospholipase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycerol metabolic process
          lipid metabolic process
          nuclease activity
          phosphoric diester hydrolase activity
          glycerophosphodiester phosphodiesterase activity
          hydrolase activity
          phosphoric ester hydrolase activity
          T/G mismatch-specific endonuclease activity
          retroviral 3' processing activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426875 NM_024228.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426875 AK003726.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426875 BC002172.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426875 NM_024228.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426875 AK003726.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426875 BC002172.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195101 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 528469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 873462 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 167711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 438491 AK009290.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 963800 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 96202 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150950 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426875 NM_024228.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426875 AK003726.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426875 BC002172.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426875 NM_024228.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426875 AK003726.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426875 BC002172.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426875 NM_024228.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426875 AK003726.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426875 BC002172.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426875 NM_024228.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426875 AK003726.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426875 BC002172.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25502081 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150950 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01630266 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01630259 - -
          Whole Mouse Genome G4122A A_51_P426875 NM_024228.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P426875 AK003726.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P426875 BC002172.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P426875 NM_024228.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P426875 AK003726.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P426875 BC002172.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134804 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150950 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06273519 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06273526 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06273527 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06273517 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06273535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25502171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06273499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06644098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06388981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06273504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06273537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06629780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06072354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06147818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06653404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06521864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06273489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06644101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06273513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25502107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06273490 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10557475 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557480 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557472 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557473 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557476 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557479 - -
          Affymetrix HT MG-430 PM Array Plate 1449526_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10557470 - -
          Mouse Genome 430 2.0 Array 1449526_a_at - -
          Mouse Genome 430 2.0 Array 1449526_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68616_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68616_at - -
          Affymetrix Mouse Clariom S Array PSR0700015491.mm.1 TC0700001815.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700015491.mm.1 NM_024228.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015491.mm.1 NM_024228.2 Refseq
          Mouse Expression Set 430 1449526_a_at - -
          Mouse Expression Set 430 1449526_PM_a_at - -
          Murine Genome U74v2 Set 104217_PM_at - -
          Murine Genome U74v2 Set 104217_at - -
          Mouse Genome 430A 2.0 Array 1449526_a_at - -
          Mouse Genome 430A 2.0 Array 1449526_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449526_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449526_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6110292 - -
          Illumina mouse-6 v1.0 expression beadchip 6110292 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006110292 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2685882 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2685882 - -
          mouse-6 v1.1 (Illumina) scl32054.9.1_17-S - -
          Illumina Sentrix mouse-6.v1 bead array 6110292 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32054.9.1_17-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6110292 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2893879 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2893879 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline