Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00470447_m1
          See other 1500015O10RIK GE Assays ›
          Gene Symbol:
          1500015O10Rik
          Gene Name:
          RIKEN cDNA 1500015O10 gene
          Gene Aliases:
          Ecrg4
          Chromosome Location:
          Chr.1: 43730602 - 43742564 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00470447_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1500015O10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024283.3 NP_077245.2 3 - 4 566 85
          GenBank mRNA AK018770.1 - 3 - 4 321 85
          AK049805.1 - 3 - 4 567 85
          BC002254.1 - 3 - 4 499 3493553 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000329.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1500015O10Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cyclin-dependent protein serine/threonine kinase by cyclin degradation
          G1 to G0 transition
          cellular senescence
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P404463 NM_024283.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P404463 AK018770.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P404463 AK049805.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P404463 BC002254.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P404463 NM_024283.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P404463 AK018770.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P404463 AK049805.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P404463 BC002254.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 206649 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 412229 AK034181.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 412229 AK047398.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 571285 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 332147 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394087 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 912998 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 77697 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 7561 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170163 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162809 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P404463 NM_024283.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P404463 AK018770.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P404463 AK049805.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P404463 BC002254.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P404463 NM_024283.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P404463 AK018770.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P404463 AK049805.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P404463 BC002254.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20190493 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04081294 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162809 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170163 - -
          Whole Mouse Genome G4122A A_51_P404463 NM_024283.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404463 AK018770.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404463 AK049805.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404463 BC002254.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404463 NM_024283.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404463 AK018770.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404463 AK049805.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404463 BC002254.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136775 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162809 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170163 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00216161 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00216166 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00216165 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00216159 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00216150 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00216155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20190550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00288333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01051025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01051019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01114405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00598065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00121756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01142506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01114394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00767307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00216150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00121742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00216156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01114395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00363874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00780321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20190574 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10345923 - -
          Affymetrix Mouse Gene 1.0 ST Array 10345924 - -
          Affymetrix Mouse Gene 1.0 ST Array 10345925 - -
          Affymetrix Mouse Gene 1.0 ST Array 10345922 - -
          Affymetrix HT MG-430 PM Array Plate 1440780_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1423261_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1460049_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10345921 - -
          Mouse Genome 430 2.0 Array 1440780_x_at - -
          Mouse Genome 430 2.0 Array 1423261_at - -
          Mouse Genome 430 2.0 Array 1460049_s_at - -
          Mouse Genome 430 2.0 Array 1460049_PM_s_at - -
          Mouse Genome 430 2.0 Array 1423261_PM_at - -
          Mouse Genome 430 2.0 Array 1440780_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78896_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78896_at - -
          Affymetrix Mouse Clariom S Array PSR0100002412.mm.1 NM_024283.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002412.mm.1 NM_024283.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002412.mm.1 TC0100000329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100002414.mm.1 NM_024283.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002414.mm.1 NM_024283.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002414.mm.1 TC0100000329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100002413.mm.1 NM_024283.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002413.mm.1 NM_024283.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002413.mm.1 TC0100000329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100002416.mm.1 NM_024283.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002416.mm.1 NM_024283.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002416.mm.1 TC0100000329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100002415.mm.1 NM_024283.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002415.mm.1 NM_024283.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100002415.mm.1 TC0100000329.mm.1 MTA TC
          Mouse Expression Set 430 1423261_PM_at - -
          Mouse Expression Set 430 1423261_at - -
          Mouse Expression Set 430 1440780_x_at - -
          Mouse Expression Set 430 1460049_s_at - -
          Mouse Expression Set 430 1440780_PM_x_at - -
          Mouse Expression Set 430 1460049_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1423261_PM_at - -
          Mouse Genome 430A 2.0 Array 1423261_at - -
          Affymetrix HT Mouse Genome 430A Array 1423261_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423261_at - -
          Affymetrix HT Mouse Genome 430B Array 1460049_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1440780_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1440780_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1460049_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 768385 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3130368 - -
          Illumina mouse-6 v1.0 expression beadchip 3130368 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003130368 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1249000 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249000 - -
          mouse-6 v1.1 (Illumina) scl18005.4.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 3130368 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl18005.4.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3130368 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1249000 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1249000 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl18005.4.1_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline