Search
Search

Klhl13| Gene Symbol: Klhl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290476.1 | NP_001277405.1 | 5 - 6 | 1537 | 73 | |
| NM_026167.4 | NP_080443.1 | 7 - 8 | 1556 | 73 | ||
| GenBank mRNA | AK004677.1 | - | 7 - 8 | 1528 | 73 | |
| AK122491.1 | - | 5 - 6 | 1537 | 73 | ||
| BC100393.1 | - | 6 - 7 | 1521 | 30737426 | 73 | |
Set Membership: |
|