Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00470808_m1
          See other RNF151 GE Assays ›
          Gene Symbol:
          Rnf151
          Gene Name:
          ring finger protein 151
          Gene Aliases:
          1700010O16Rik, AV047371
          Chromosome Location:
          Chr.17: 24715839 - 24718057 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00470808_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf151
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026205.3 NP_080481.1 2 - 3 220 86
          GenBank mRNA AK005850.1 - 2 - 3 181 86
          AK005854.1 - 2 - 3 176 86
          BC049562.1 - 2 - 3 220 6774166 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001639.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf151

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatogenesis
          protein ubiquitination
          cell differentiation
          ubiquitin-protein transferase activity
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099197 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099198 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2093211 AK005854.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2093211 BC049562.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2093211 AK005850.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2093211 NM_026205.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471116 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 147751 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118370 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2093211 AK005854.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2093211 BC049562.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2093211 AK005850.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2093211 NM_026205.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099170 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118370 - -
          Whole Mouse Genome G4122A A_51_P405314 BC049562.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P405314 AK005850.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P405314 NM_026205.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113748 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101328 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118370 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13523539 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13408539 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13136053 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13136048 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13408533 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13136054 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13566074 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13408538 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099198 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13523540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13408546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13408533 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10448735 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448732 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448736 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448733 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448734 - -
          Affymetrix HT MG-430 PM Array Plate 1451009_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10448731 - -
          Mouse Genome 430 2.0 Array 1451009_PM_at - -
          Mouse Genome 430 2.0 Array 1451009_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67504_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67504_at - -
          Affymetrix Mouse Clariom S Array PSR1700014696.mm.1 NM_026205.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014696.mm.1 NM_026205.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014696.mm.1 TC1700001639.mm.1 MTA TC
          Mouse Expression Set 430 1451009_at - -
          Mouse Expression Set 430 1451009_PM_at - -
          Murine Genome U74v2 Set 165486_PM_i_at - -
          Murine Genome U74v2 Set 165486_i_at - -
          Murine Genome U74v2 Set 169308_at - -
          Murine Genome U74v2 Set 169308_PM_at - -
          Mouse Genome 430A 2.0 Array 1451009_PM_at - -
          Mouse Genome 430A 2.0 Array 1451009_at - -
          Affymetrix HT Mouse Genome 430A Array 1451009_at - -
          Affymetrix HT Mouse Genome 430A Array 1451009_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 663168 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 610692 - -
          Illumina mouse-6 v1.0 expression beadchip 610692 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610692 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2648122 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2648122 - -
          mouse-6 v1.1 (Illumina) scl50200.5.1_98-S - -
          Illumina Sentrix mouse-6.v1 bead array 610692 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50200.5.1_98-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610692 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2648122 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2648122 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline