Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00470868_s1
          See other H2AFY3 GE Assays ›
          Gene Symbol:
          H2afy3
          Gene Name:
          H2A histone family, member Y3
          Gene Aliases:
          4933432H23Rik
          Chromosome Location:
          Chr.15: 62217541 - 62219451 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00470868_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H2afy3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003523.1 1 1299 73
          GenBank mRNA AK017022.1 - 1 1251 73
          AK017051.1 - 1 1295 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001612.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          histone

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487668 NR_003523.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487668 AK017022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487668 NR_003523.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487668 AK017022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487668 NR_003523.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487668 AK017022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487668 NR_003523.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487668 AK017022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487668 NR_003523.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487668 AK017022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487668 NR_003523.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487668 AK017022.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12357618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12357629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12435616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12438702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12438704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12357624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30269033 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10428935 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428934 - -
          Affymetrix HT MG-430 PM Array Plate 1421613_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1450307_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10428933 - -
          Mouse Genome 430 2.0 Array 1421613_PM_at - -
          Mouse Genome 430 2.0 Array 1421613_at - -
          Mouse Genome 430 2.0 Array 1450307_PM_x_at - -
          Mouse Genome 430 2.0 Array 1450307_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67552_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67552_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500012082.mm.1 NR_003523.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012082.mm.1 TC1500001612.mm.1 MTA TC
          Mouse Expression Set 430 1421613_at - -
          Mouse Expression Set 430 1421613_PM_at - -
          Mouse Expression Set 430 1450307_PM_x_at - -
          Mouse Expression Set 430 1421612_PM_a_at - -
          Mouse Expression Set 430 1450307_x_at - -
          Mouse Expression Set 430 1421612_a_at - -
          Mouse Genome 430A 2.0 Array 1450307_x_at - -
          Mouse Genome 430A 2.0 Array 1421612_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1450307_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1421613_at - -
          Mouse Genome 430A 2.0 Array 1421613_PM_at - -
          Mouse Genome 430A 2.0 Array 1421612_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421613_at - -
          Affymetrix HT Mouse Genome 430A Array 1450307_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1450307_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1421613_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 745619 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2741360 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2741360 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2741360 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline