| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10491330 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1420580_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10491329 |
- |
- |
| Mouse Genome 430 2.0 Array |
1420580_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1420580_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
67576_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
67576_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0300001219.mm.1 |
NR_027966.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001219.mm.1 |
NR_027966.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001219.mm.1 |
TC0300000220.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001220.mm.1 |
NR_027966.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001220.mm.1 |
NR_027966.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001220.mm.1 |
TC0300000220.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1420580_at |
- |
- |
| Mouse Expression Set 430 |
1420580_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1420580_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1420580_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1420580_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1420580_PM_at |
- |
- |