Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00471003_s1
          See other FIBIN GE Assays ›
          Gene Symbol:
          Fibin
          Gene Name:
          fin bud initiation factor homolog (zebrafish)
          Gene Aliases:
          1110018M03Rik
          Chromosome Location:
          Chr.2: 110360925 - 110362993 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00471003_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fibin
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026271.1 NP_080547.1 1 356 73
          GenBank mRNA AB236893.1 - 1 158 73
          AK003790.1 - 1 356 73
          AK011548.1 - 1 527 73
          AK012026.1 - 1 539 73
          AK159648.1 - 1 489 73
          BC027250.1 - 1 345 3603588 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004309.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fibin

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P283473 AK159648.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P283473 AK003790.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P283473 NM_026271.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P283473 AK159648.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P283473 AK003790.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P283473 NM_026271.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 578835 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148208 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P283473 AK159648.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P283473 AK003790.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P283473 NM_026271.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P283473 AK159648.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P283473 AK003790.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P283473 NM_026271.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21520290 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148208 - -
          Whole Mouse Genome G4122A A_51_P283473 AK159648.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P283473 AK003790.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P283473 NM_026271.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P283473 AK159648.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P283473 AK003790.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P283473 NM_026271.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119529 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148208 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01420106 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02222319 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02222317 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01420085 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01420105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21520361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02182773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01420082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02207049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01712793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01455945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01479075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01420123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02185947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01479069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01455944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02219784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01712797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01741407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21520269 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10485713 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485712 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485715 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485714 - -
          Affymetrix HT MG-430 PM Array Plate 1419376_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10485711 - -
          Mouse Genome 430 2.0 Array 1419376_at - -
          Mouse Genome 430 2.0 Array 1419376_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67606_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67606_at - -
          Affymetrix Mouse Clariom S Array PSR0200036086.mm.1 NM_026271.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036086.mm.1 NM_026271.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036086.mm.1 TC0200004309.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200036088.mm.1 NM_026271.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036088.mm.1 NM_026271.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036088.mm.1 TC0200004309.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200036087.mm.1 NM_026271.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036087.mm.1 NM_026271.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036087.mm.1 TC0200004309.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200036085.mm.1 NM_026271.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036085.mm.1 NM_026271.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036085.mm.1 TC0200004309.mm.1 MTA TC
          Mouse Expression Set 430 1419376_at - -
          Mouse Expression Set 430 1419376_PM_at - -
          Murine Genome U74v2 Set 169782_PM_r_at - -
          Murine Genome U74v2 Set 169782_r_at - -
          Mouse Genome 430A 2.0 Array 1419376_at - -
          Mouse Genome 430A 2.0 Array 1419376_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419376_at - -
          Affymetrix HT Mouse Genome 430A Array 1419376_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 770980 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 70441 - -
          Illumina mouse-6 v1.0 expression beadchip 70441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000070441 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2698637 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2698637 - -
          mouse-6 v1.1 (Illumina) scl18885.1.184_77-S - -
          Illumina Sentrix mouse-6.v1 bead array 70441 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl18885.1.184_77-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 70441 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2698637 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2698637 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0