Search
Search

Polr2g| Gene Symbol: Polr2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026329.2 | NP_080605.1 | 3 - 4 | 371 | 69 | |
| GenBank mRNA | AK010687.1 | - | 3 - 4 | 340 | 69 | |
| AK011627.1 | - | 3 - 4 | 372 | 69 | ||
| AK013134.1 | - | 3 - 4 | 342 | 69 | ||
| AK017473.1 | - | 3 - 4 | 350 | 69 | ||
| BC005580.1 | - | 3 - 4 | 390 | 3968561 | 69 | |
| BC055278.1 | - | 3 - 4 | 321 | 5699685 | 69 | |
Set Membership: |
|