Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00471239_m1
          See other JAGN1 GE Assays ›
          Gene Symbol:
          Jagn1
          Gene Name:
          jagunal homolog 1 (Drosophila)
          Gene Aliases:
          5830427H10Rik
          Chromosome Location:
          Chr.6: 113442517 - 113448229 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00471239_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Jagn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026365.3 NP_080641.1 1 - 2 228 69
          GenBank mRNA AK003673.1 - 1 - 2 154 69
          AK017953.1 - 1 - 2 163 69
          AK141417.1 - 1 - 2 208 69
          AK151252.1 - 1 - 2 190 69
          BC011296.1 - 1 - 2 195 3962988 69
          BC025910.1 - 1 - 2 182 5252891 69
          BC037019.1 - 1 - 2 182 5364053 69
          BU609459.1 - 1 - 2 229 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001250.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Jagn1

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          neutrophil mediated immunity
          transport
          endoplasmic reticulum organization
          protein transport
          vesicle-mediated transport
          neutrophil differentiation
          granulocyte colony-stimulating factor signaling pathway
          defense response to fungus
          neutrophil migration
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24759516 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24759515 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150608 AK170345.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150608 AK151252.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150608 BC011296.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150608 BC025910.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150608 BC037019.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150608 AK170345.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150608 AK151252.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150608 BC011296.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150608 BC025910.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150608 BC037019.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444982 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481403 AK033068.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481403 NM_001081424.1 refseq_mrna_mouse_NM
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153515 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150608 AK170345.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150608 AK151252.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150608 BC011296.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150608 BC025910.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150608 BC037019.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150608 AK170345.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150608 AK151252.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150608 BC011296.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150608 BC025910.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150608 BC037019.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24759519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24759520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24759515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24759517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24759516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24759522 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06031022 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153515 - -
          Whole Mouse Genome G4122A A_52_P663384 AK034671.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P663384 AK043698.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150608 AK170345.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150608 AK151252.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150608 BC011296.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150608 BC025910.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150608 BC037019.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150608 AK170345.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150608 AK151252.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150608 BC011296.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150608 BC025910.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150608 BC037019.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124868 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153515 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05211425 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24759516 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24759515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24759572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24759544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24759536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24759520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05048199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24759541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05745072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24759506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05530797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05211429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05745071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24759534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05799317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24759500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24759535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05211425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24759499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05211426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05050178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24759545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05050174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05229260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05799314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24759504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24759531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24759538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24759522 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10540677 - -
          Affymetrix Mouse Gene 1.0 ST Array 10540678 - -
          Affymetrix HT MG-430 PM Array Plate 1417064_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10540676 - -
          Mouse Genome 430 2.0 Array 1417064_PM_at - -
          Mouse Genome 430 2.0 Array 1417064_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67767_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67767_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600009728.mm.1 TC0600001250.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600009728.mm.1 NM_001205025.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009728.mm.1 NM_001205025.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009728.mm.1 NM_001243739.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009728.mm.1 NM_001243739.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009728.mm.1 NM_026365.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009728.mm.1 NM_026365.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009722.mm.1 TC0600001250.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600009722.mm.1 NM_001205025.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009722.mm.1 NM_001205025.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009722.mm.1 NM_001243739.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009722.mm.1 NM_001243739.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009722.mm.1 NM_026365.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009722.mm.1 NM_026365.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009726.mm.1 TC0600001250.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600009726.mm.1 NM_001243739.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009726.mm.1 NM_001243739.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009726.mm.1 NM_026365.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009726.mm.1 NM_026365.3 Refseq
          Mouse Expression Set 430 1417064_at - -
          Mouse Expression Set 430 1417064_PM_at - -
          Murine Genome U74v2 Set 93579_at - -
          Murine Genome U74v2 Set 93579_PM_at - -
          Mouse Genome 430A 2.0 Array 1417064_PM_at - -
          Mouse Genome 430A 2.0 Array 1417064_at - -
          Affymetrix HT Mouse Genome 430A Array 1417064_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417064_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 926268 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105890161 - -
          Illumina mouse-6 v1.0 expression beadchip 105890161 - -
          Illumina mouse-6 v1.0 expression beadchip 1690093 - -
          Illumina mouse-6 v1.0 expression beadchip 101660711 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001690093 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221859 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220712 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221859 - -
          mouse-6 v1.1 (Illumina) scl29632.2_358-S - -
          Illumina Sentrix mouse-6.v1 bead array 1690093 - -
          Illumina Sentrix mouse-6.v1 bead array 101660711 - -
          Illumina Sentrix mouse-6.v1 bead array 105890161 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29632.2_358-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105890161 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1690093 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101660711 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1221859 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221859 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1217553 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220712 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl29632.2_358-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline