Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00471507_m1
          See other MURC GE Assays ›
          Gene Symbol:
          Murc
          Gene Name:
          muscle-related coiled-coil protein
          Gene Aliases:
          2310039E09Rik
          Chromosome Location:
          Chr.4: 48663514 - 48673492 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00471507_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Murc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026509.3 NP_080785.2 1 - 2 523 116
          GenBank mRNA AK009691.1 - 1 - 2 480 116
          BC089601.1 - 1 - 2 523 6788090 116
          EU487254.1 - 1 - 2 415 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000536.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Murc

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          Rho protein signal transduction
          multicellular organism development
          muscle organ development
          regulation of gene expression
          cell differentiation
          positive regulation of GTPase activity
          positive regulation of transcription from RNA polymerase II promoter
          cardiac myofibril assembly
          GTPase activator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954331 BC089601.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954331 NM_026509.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954331 AK009691.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121107 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 895136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973376 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 522392 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136018 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954331 BC089601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954331 NM_026509.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954331 AK009691.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22901448 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136018 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00867742 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05230129 - -
          Whole Mouse Genome G4122A A_51_P378029 BC089601.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P378029 NM_026509.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P378029 AK009691.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133042 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136018 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03671370 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03671364 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03671371 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03671373 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03671368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22901447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03671370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03538559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03329786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03619179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03329797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03989987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03538569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03580953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03538567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03671371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03671362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03671368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03329792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03538574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03671358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03270946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03671357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03538570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04051564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03460134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03329800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03671373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03671367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22901469 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10504904 - -
          Affymetrix Mouse Gene 1.0 ST Array 10504903 - -
          Affymetrix HT MG-430 PM Array Plate 1420551_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10504902 - -
          Mouse Genome 430 2.0 Array 1420551_PM_at - -
          Mouse Genome 430 2.0 Array 1420551_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68016_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68016_at - -
          Affymetrix Mouse Clariom S Array PSR0400003993.mm.1 NM_026509.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003993.mm.1 NM_026509.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003993.mm.1 TC0400000536.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400003992.mm.1 NM_026509.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003992.mm.1 NM_026509.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003992.mm.1 TC0400000536.mm.1 MTA TC
          Mouse Expression Set 430 1420551_at - -
          Mouse Expression Set 430 1420551_PM_at - -
          Mouse Genome 430A 2.0 Array 1420551_at - -
          Mouse Genome 430A 2.0 Array 1420551_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420551_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420551_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 558905 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3870068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003870068 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228951 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228951 - -
          mouse-6 v1.1 (Illumina) scl25446.2_385-S - -
          Illumina Sentrix mouse-6.v1 bead array 3870068 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25446.2_385-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3870068 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2897214 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2897214 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline