Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00471518_m1
          See other EMC4 GE Assays ›
          Gene Symbol:
          Emc4
          Gene Name:
          ER membrane protein complex subunit 4
          Gene Aliases:
          2610318K02Rik, Tmem85
          Chromosome Location:
          Chr.2: 112363019 - 112368027 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00471518_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Emc4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026519.3 NP_080795.1 2 - 3 324 56
          GenBank mRNA AK003668.1 - 2 - 3 292 56
          AK012047.1 - 2 - 3 325 56
          AK078165.1 - 2 - 3 325 56
          AK167745.1 - 2 - 3 276 56
          BC051926.1 - 2 - 3 280 5710720 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004352.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Emc4

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21528513 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21528514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21528510 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21528511 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P215066 AK167745.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P215066 BC051926.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P215066 NM_026519.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P215066 AK003668.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P215066 AK012047.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P215066 AK167745.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P215066 BC051926.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P215066 NM_026519.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P215066 AK003668.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P215066 AK012047.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 232629 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 230924 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 213901 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 3120 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172834 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P215066 AK167745.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P215066 BC051926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P215066 NM_026519.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P215066 AK003668.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P215066 AK012047.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P215066 AK167745.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P215066 BC051926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P215066 NM_026519.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P215066 AK003668.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P215066 AK012047.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21528532 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172834 - -
          Whole Mouse Genome G4122A A_51_P215066 AK167745.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P215066 BC051926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P215066 NM_026519.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P215066 AK003668.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P215066 AK012047.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P215066 AK167745.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P215066 BC051926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P215066 NM_026519.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P215066 AK003668.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P215066 AK012047.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129505 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172834 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02198437 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01627906 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01693977 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01693980 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01627909 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01693979 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21528513 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21528511 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21528510 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21528514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21528493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01653661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01693973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01836023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01627909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01790129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01693975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01157787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01835818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21528502 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10485831 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485835 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485834 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485832 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485833 - -
          Affymetrix HT MG-430 PM Array Plate 1438369_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1418124_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10485830 - -
          Mouse Genome 430 2.0 Array 1438369_PM_x_at - -
          Mouse Genome 430 2.0 Array 1438369_x_at - -
          Mouse Genome 430 2.0 Array 1418124_at - -
          Mouse Genome 430 2.0 Array 1458916_at - -
          Mouse Genome 430 2.0 Array 1458916_PM_at - -
          Mouse Genome 430 2.0 Array 1418124_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68032_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68032_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200036278.mm.1 NM_026519.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036278.mm.1 NM_026519.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036278.mm.1 TC0200004352.mm.1 MTA TC
          Mouse Expression Set 430 1418124_PM_at - -
          Mouse Expression Set 430 1438369_x_at - -
          Mouse Expression Set 430 1438369_PM_x_at - -
          Mouse Expression Set 430 1418124_at - -
          Murine Genome U74v2 Set 94035_PM_at - -
          Murine Genome U74v2 Set 94035_at - -
          Mouse Genome 430A 2.0 Array 1418124_PM_at - -
          Mouse Genome 430A 2.0 Array 1438369_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1438369_x_at - -
          Mouse Genome 430A 2.0 Array 1418124_at - -
          Affymetrix HT Mouse Genome 430A Array 1438369_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1438369_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1418124_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418124_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 900178 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5910148 - -
          Illumina mouse-6 v1.0 expression beadchip 5910148 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005910148 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2657163 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2657163 - -
          mouse-6 v1.1 (Illumina) scl18868.1.64_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 5910148 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl18868.1.64_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5910148 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2809247 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2809239 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2809239 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2809247 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl18868.1.64_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline