| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106819 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106913 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106816 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106917 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106876 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106873 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106818 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106870 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106916 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106872 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106915 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106875 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106874 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106871 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P106527 |
BC024332.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P106527 |
NM_026633.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P106527 |
AK003564.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P106527 |
AK020614.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P106527 |
BC024332.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P106527 |
NM_026633.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P106527 |
AK003564.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P106527 |
AK020614.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
517575 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
448123 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829828 |
BC040756.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829828 |
XM_001476619.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829828 |
XM_001480054.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
552731 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
122254 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P158296 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P106527 |
BC024332.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P106527 |
NM_026633.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P106527 |
AK003564.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P106527 |
AK020614.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P106527 |
BC024332.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P106527 |
NM_026633.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P106527 |
AK003564.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P106527 |
AK020614.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P106527 |
BC024332.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P106527 |
NM_026633.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P106527 |
AK003564.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P106527 |
AK020614.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P106527 |
BC024332.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P106527 |
NM_026633.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P106527 |
AK003564.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P106527 |
AK020614.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106866 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106871 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106855 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106909 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106917 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106879 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106904 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106915 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106913 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106910 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106872 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106876 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106867 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106907 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106869 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106912 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106900 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106861 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106897 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106878 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106863 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106896 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106881 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106893 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106911 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106906 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106898 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106902 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106891 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106858 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106856 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106875 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106899 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31106857 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P158296 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P106527 |
BC024332.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P106527 |
NM_026633.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P106527 |
AK003564.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P106527 |
AK020614.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P106527 |
BC024332.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P106527 |
NM_026633.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P106527 |
AK003564.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P106527 |
AK020614.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P127659 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P158296 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13175336 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13073642 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13175326 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13175328 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13175330 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13073655 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13073635 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13073641 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13073648 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106876 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106873 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106915 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106916 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106818 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106913 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106871 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106870 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106875 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106819 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106917 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106816 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106872 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31106874 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106898 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106769 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106770 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106774 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106822 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106827 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106941 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106893 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106802 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106849 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106763 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106839 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106872 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106810 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106942 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106902 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106782 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106787 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106855 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106932 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106801 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106879 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106767 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106773 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106853 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106920 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106925 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106912 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106866 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106778 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106869 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106852 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106777 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106847 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106804 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106818 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106789 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106922 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106858 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106816 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106794 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106915 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106765 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106934 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106861 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106762 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106831 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106917 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106825 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106817 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106929 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106899 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106814 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106907 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106918 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106809 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106911 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106780 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106830 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106936 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106781 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106876 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106812 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106784 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106836 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106833 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106938 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106826 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106764 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106857 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106875 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106842 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106896 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106909 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106800 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106927 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106798 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106790 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106913 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106771 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106889 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106837 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106796 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106803 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106806 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106820 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106900 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31106891 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13175332 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106847 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106818 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106777 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106789 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13511722 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106857 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13103103 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106802 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106855 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106786 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106823 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106824 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106879 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106809 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106849 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106853 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106800 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106876 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106781 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13409526 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13103107 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106790 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106873 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106783 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106825 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13319163 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106804 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13175317 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106830 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13131666 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106776 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106774 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106771 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106858 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106778 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106810 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106820 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106792 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106798 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13175328 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106852 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106773 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106839 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106889 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106827 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13103117 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106875 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106787 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13409511 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106850 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106812 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106806 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106784 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13175319 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13103127 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106833 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106794 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106780 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106866 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106796 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106822 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106842 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13319165 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31106813 |
- |
- |