Search
Search

Fam114a1| Gene Symbol: Fam114a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026667.3 | NP_080943.3 | 5 - 6 | 735 | 63 | |
| GenBank mRNA | AK007950.1 | - | 5 - 6 | 693 | 63 | |
| AK014725.1 | - | 5 - 6 | 705 | 63 | ||
| AK017336.1 | - | 5 - 6 | 695 | 63 | ||
| AK020262.1 | - | 5 - 6 | 696 | 63 | ||
| AK030360.1 | - | 5 - 6 | 736 | 63 | ||
| AM162548.1 | - | 5 - 6 | 739 | 63 | ||
| BC027235.1 | - | 5 - 6 | 698 | 3603083 | 63 | |
| BC110359.1 | - | 5 - 6 | 736 | 5325099 | 63 | |
Set Membership: |
|