Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00472039_s1
          See other KRTAP28-13 GE Assays ›
          Gene Symbol:
          Krtap28-13
          Gene Name:
          keratin associated protein 28-13
          Gene Aliases:
          5530401N06Rik
          Chromosome Location:
          Chr.1: 83060936 - 83061915 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00472039_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krtap28-13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK017438.1 - 1 652 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P269721 AK017438.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P269721 AK017438.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P269721 AK017438.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P269721 AK017438.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154994 AV294026.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269721 AK017438.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P269721 AK017438.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00798032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20396376 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1422298_PM_at - -
          Mouse Genome 430 2.0 Array 1422298_PM_at - -
          Mouse Genome 430 2.0 Array 1422298_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71386_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71386_PM_at - -
          Mouse Expression Set 430 1422298_PM_at - -
          Mouse Expression Set 430 1422298_at - -
          Mouse Genome 430A 2.0 Array 1422298_PM_at - -
          Mouse Genome 430A 2.0 Array 1422298_at - -
          Affymetrix HT Mouse Genome 430A Array 1422298_at - -
          Affymetrix HT Mouse Genome 430A Array 1422298_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 635199 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2523891 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0