Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00472247_g1
          See other B3GNT3 GE Assays ›
          Gene Symbol:
          B3gnt3
          Gene Name:
          UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3
          Gene Aliases:
          2210008L19Rik
          Chromosome Location:
          Chr.8: 71691373 - 71702627 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00472247_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B3gnt3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028189.3 NP_082465.3 2 - 3 668 84
          GenBank mRNA AK008674.1 - 707 84
          AK144117.1 - 658 84
          AK144180.1 - 2 - 3 640 84
          AY037785.1 - 637 84
          AY039028.1 - 1 - 2 587 84
          BC026418.1 - 2 - 3 660 4218953 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002440.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene B3gnt3

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          poly-N-acetyllactosamine biosynthetic process
          galactosyltransferase activity
          N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups
          beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,3-N-acetylglucosaminyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958414 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958357 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958416 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958363 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958356 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958360 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958364 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958362 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958358 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958417 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958415 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958365 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139546 NM_028189.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 158695 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 518962 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298986 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432284 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 182080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 44458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 285102 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 153804 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173754 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106316 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139546 NM_028189.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P240760 AY037785.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P240760 AY039028.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P240760 NM_028189.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P240760 AK008674.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25958348 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173754 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01764872 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106316 - -
          Whole Mouse Genome G4122A A_51_P240760 AY037785.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240760 AY039028.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240760 NM_028189.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240760 AK008674.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126785 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173754 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106316 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06930180 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06930178 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07116870 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07116879 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07116875 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958363 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958357 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958358 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958415 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958362 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958417 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958365 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958356 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958364 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958416 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958360 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25958414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25958290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07372061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06971116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07145102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07252026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06971127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07379521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06895793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06686913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06867128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07145100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06930179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06971115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07429249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06895791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06686915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06930171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06709920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06935828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25958417 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10579604 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579603 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579605 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579606 - -
          Affymetrix HT MG-430 PM Array Plate 1422211_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10579602 - -
          Mouse Genome 430 2.0 Array 1422211_a_at - -
          Mouse Genome 430 2.0 Array 1422211_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72297_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72297_at - -
          Affymetrix Mouse Clariom S Array PSR0800018659.mm.1 NM_028189.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800018659.mm.1 NM_028189.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800018659.mm.1 TC0800002440.mm.1 MTA TC
          Mouse Expression Set 430 1422211_a_at - -
          Mouse Expression Set 430 1422211_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1422211_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1422211_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1422211_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1422211_a_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2850471 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002850471 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2776500 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2776500 - -
          mouse-6 v1.1 (Illumina) scl34666.4_288-S - -
          Illumina Sentrix mouse-6.v1 bead array 2850471 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34666.4_288-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2850471 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2907972 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2776503 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2907972 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline