| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10523892 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1422840_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10523891 |
- |
- |
| Mouse Genome 430 2.0 Array |
1422840_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1422840_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
73318_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
73318_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0500009172.mm.1 |
NM_001276397.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009172.mm.1 |
TC0500001085.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1422840_PM_at |
- |
- |
| Mouse Expression Set 430 |
1422840_at |
- |
- |
| Murine Genome U74v2 Set |
167057_i_at |
- |
- |
| Murine Genome U74v2 Set |
167057_PM_i_at |
- |
- |
| Murine Genome U74v2 Set |
169973_PM_at |
- |
- |
| Murine Genome U74v2 Set |
169973_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1422840_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1422840_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1422840_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1422840_at |
- |
- |