Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00472307_g1
          See other LOC100043315 GE Assays ›
          Gene Symbol:
          LOC100043315
          Gene Name:
          uncharacterized LOC100043315
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 95097622 - 95149318 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00472307_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100043315
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC106998.1 - 1 - 2 246 139

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2175332 BC115946.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2175332 BC127134.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2175332 BC106998.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2175332 BC106999.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2175332 BC114343.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136165 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12212631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12119796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11874742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12119784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12451189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456399 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12128320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12128309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12119791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11958475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12151949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12165878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12128312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12119792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11848233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30456422 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10431837 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431838 - -
          Affymetrix HT MG-430 PM Array Plate 1450328_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10431836 - -
          Mouse Genome 430 2.0 Array 1450328_at - -
          Mouse Genome 430 2.0 Array 1450328_PM_at - -
          Mouse Expression Set 430 1450328_PM_at - -
          Mouse Expression Set 430 1450328_at - -
          Mouse Genome 430A 2.0 Array 1450328_PM_at - -
          Mouse Genome 430A 2.0 Array 1450328_at - -
          Affymetrix HT Mouse Genome 430A Array 1450328_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450328_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline