Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00472505_g1
          See other VMN2R-PS54 GE Assays ›
          Gene Symbol:
          Vmn2r-ps54
          Gene Name:
          vomeronasal 2, receptor, pseudogene 54
          Gene Aliases:
          1700108E19Rik, 4930562N12Rik, Casr-rs5, ENSMUSG00000078527, Gm10948, Gprc2a-rs5
          Chromosome Location:
          Chr.7: 41668014 - 41727193 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00472505_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn2r-ps54
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_004441.1 2 - 3 210 139
          GenBank mRNA AK016194.1 - 2 - 3 244 139
          AK030272.1 - 2 - 3 211 139

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000734.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK016194.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002135 AK030272.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002135 NR_004441.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685148 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 918788 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904661 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 364699 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 605244 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 873774 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 910798 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 304300 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727963 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 239465 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 229102 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 565172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 658176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 9915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 67821 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82528 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142154 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 22212 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002135 AK030272.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002135 NR_004441.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06175444 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06175436 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140721 - -
          Whole Mouse Genome G4122A A_51_P276681 AK016194.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276681 AK030272.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276681 NR_004441.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276681 AK016194.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276681 AK030272.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276681 NR_004441.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109695 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113927 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06418940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05980465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06641543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06242639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06418939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06242642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06131640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06530594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06242650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06242647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06242638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06196278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06131632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06418943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06131639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25078190 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552334 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552342 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536080 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562642 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552332 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536081 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536084 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536082 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552333 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552336 - -
          Affymetrix HT MG-430 PM Array Plate 1430504_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1450274_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10536079 - -
          Affymetrix Mouse Gene 1.1 ST Array 10536083 - -
          Affymetrix Mouse Gene 1.1 ST Array 10552331 - -
          Affymetrix Mouse Gene 1.1 ST Array 10552335 - -
          Affymetrix Mouse Gene 1.1 ST Array 10562641 - -
          Mouse Genome 430 2.0 Array 1430504_at - -
          Mouse Genome 430 2.0 Array 1450274_PM_at - -
          Mouse Genome 430 2.0 Array 1450274_at - -
          Mouse Genome 430 2.0 Array 1430504_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75835_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75835_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700005973.mm.1 TC0700000734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005973.mm.1 NR_004441.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005974.mm.1 TC0700000734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005974.mm.1 NR_004441.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005975.mm.1 TC0700000734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005975.mm.1 NR_004441.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005990.mm.1 TC0700000734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005990.mm.1 NR_004441.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005979.mm.1 TC0700000734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005979.mm.1 NR_004441.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005972.mm.1 TC0700000734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005972.mm.1 NR_004441.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005976.mm.1 TC0700000734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005976.mm.1 NR_004441.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005971.mm.1 TC0700000734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005971.mm.1 NR_004441.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005978.mm.1 TC0700000734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005978.mm.1 NR_004441.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005970.mm.1 TC0700000734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005970.mm.1 NR_004441.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005983.mm.1 TC0700000734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005983.mm.1 NR_004441.1 Refseq
          Mouse Expression Set 430 1450274_at - -
          Mouse Expression Set 430 1450274_PM_at - -
          Mouse Expression Set 430 1430504_at - -
          Mouse Expression Set 430 1430504_PM_at - -
          Mouse Genome 430A 2.0 Array 1450274_at - -
          Mouse Genome 430A 2.0 Array 1450274_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450274_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450274_at - -
          Affymetrix HT Mouse Genome 430B Array 1430504_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430504_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 337308 - -
          ABI Mouse Genome Survey Microarray 805191 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5670086 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226518 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2764353 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2764353 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226518 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226518 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2765653 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2765652 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline