Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00472868_m1
          See other ABO GE Assays ›
          Gene Symbol:
          Abo
          Gene Name:
          ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase, transferase B, alpha 1-3-galactosyltransferase)
          Gene Aliases:
          NAGAT
          Chromosome Location:
          Chr.2: 26842496 - 26864995 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00472868_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abo
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290444.1 NP_001277373.1 3 - 4 253 84
          NM_030718.5 NP_109643.3 3 - 4 253 84
          GenBank mRNA AB041039.1 - 2 - 3 141 84
          AK033786.1 - 3 - 4 174 84
          AK035261.1 - 3 - 4 238 84
          BC116759.1 - 3 - 4 202 40126078 84
          BC116761.1 - 3 - 4 202 40126080 84
          DV046627.1 - 3 - 4 272 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003201.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abo

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          alpha-1,3-galactosyltransferase activity
          glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase activity
          fucosylgalactoside 3-alpha-galactosyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups
          transferase activity, transferring hexosyl groups
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2037977 NM_030718.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2037977 AK035261.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 615179 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 538492 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568075 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63670 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 279790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 247325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 353262 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 99213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 943893 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 64079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 131909 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 370850 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 350740 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 100397 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11222 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106146 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167502 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2037977 NM_030718.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2037977 AK035261.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P241499 BC116759.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P241499 BC116761.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P241499 NM_030718.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P241499 AB041039.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P241499 AK033786.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21080927 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04461415 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04461390 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167502 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106146 - -
          Whole Mouse Genome G4122A A_51_P241499 BC116759.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P241499 BC116761.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P241499 NM_030718.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P241499 AB041039.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P241499 AK033786.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112086 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136913 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106146 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167502 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01631867 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01631866 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01631859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21080925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01396149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01277062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01734628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01631856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01968983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01631858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01631859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01879616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01734627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01239193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01631866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01277061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21080898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01702439 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10481119 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481116 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481121 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481120 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481118 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481115 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481117 - -
          Affymetrix HT MG-430 PM Array Plate 1421559_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10481114 - -
          Mouse Genome 430 2.0 Array 1421559_at - -
          Mouse Genome 430 2.0 Array 1421559_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80908_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80908_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200026873.mm.1 NM_030718.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026873.mm.1 TC0200003201.mm.1 MTA TC
          Mouse Expression Set 430 1421559_PM_at - -
          Mouse Expression Set 430 1421559_at - -
          Murine Genome U74v2 Set 169699_PM_at - -
          Murine Genome U74v2 Set 169699_at - -
          Mouse Genome 430A 2.0 Array 1421559_PM_at - -
          Mouse Genome 430A 2.0 Array 1421559_at - -
          Affymetrix HT Mouse Genome 430A Array 1421559_at - -
          Affymetrix HT Mouse Genome 430A Array 1421559_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 665126 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4760114 - -
          Illumina mouse-6 v1.0 expression beadchip 4760114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004760114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2670848 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2670848 - -
          mouse-6 v1.1 (Illumina) scl19518.6.1_39-S - -
          Illumina Sentrix mouse-6.v1 bead array 4760114 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19518.6.1_39-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4760114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2824780 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2824783 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2824780 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2824783 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline