Search
Search

Tbl1xr1| Gene Symbol: Tbl1xr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030732.3 | NP_109657.2 | 3 - 4 | 471 | 115 | |
| GenBank mRNA | AF268195.1 | - | 3 - 4 | 252 | 115 | |
| AK029595.1 | - | 3 - 4 | 270 | 115 | ||
| AK033347.1 | - | 3 - 4 | 341 | 115 | ||
| AK036064.1 | - | 4 - 5 | 582 | 115 | ||
| AK136255.1 | - | 3 - 4 | 293 | 115 | ||
Set Membership: |
|