| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28550067 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28550068 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28550064 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28550069 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28550066 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28550065 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28550070 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2058903 |
BC037592.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2058903 |
AK154116.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2058903 |
BC026814.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2058903 |
NM_030750.3 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
126922 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
936415 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
910715 |
AK018791.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
910715 |
AK035312.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
910715 |
AK046034.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
910715 |
AK079358.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
910715 |
AK079576.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
910715 |
AY043488.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
910715 |
BC019742.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
910715 |
NM_053267.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
533107 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
439622 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807221 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
409984 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
568081 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
856627 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
64001 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
753201 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
348281 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
651691 |
AK040668.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
211399 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
856193 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
168407 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
161774 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
77672 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P158268 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P168875 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2058903 |
BC037592.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2058903 |
AK154116.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2058903 |
BC026814.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2058903 |
NM_030750.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P42221 |
BC037592.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P42221 |
AK154116.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P42221 |
BC026814.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P42221 |
NM_030750.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P258354 |
AK043586.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550097 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550080 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550062 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550067 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550077 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550079 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550060 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550053 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550075 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550082 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550065 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550069 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550059 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550095 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550049 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550058 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550052 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550088 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550046 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550047 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550064 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550091 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550078 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550087 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550071 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550084 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550094 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550081 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550073 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550093 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28550056 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07669477 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P168875 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02550766 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P158268 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07669493 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P258354 |
AK043586.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P42221 |
BC037592.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P42221 |
AK154116.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P42221 |
BC026814.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P42221 |
NM_030750.3 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P131680 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P158268 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P168875 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10040322 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10401125 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10401129 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10401126 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28550066 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28550064 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28550067 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28550069 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28550065 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28550070 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28550068 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550075 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549989 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550002 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550056 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549986 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550010 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550033 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550023 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550067 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549946 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549938 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549954 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550053 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549912 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550062 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550079 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550028 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549987 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550047 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549974 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550064 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549958 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549917 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549998 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550037 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550001 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550080 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549930 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549948 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550031 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549969 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550021 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550025 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549914 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549993 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550077 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549994 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550068 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549982 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549944 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550035 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550072 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549975 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550078 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550110 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549934 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549967 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550094 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549922 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550052 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549932 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550091 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550059 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550104 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549995 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549933 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549939 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549996 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549957 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550084 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550082 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550004 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549907 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550006 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550046 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550042 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549918 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550069 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549945 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550049 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549915 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549928 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549924 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549999 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549965 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550008 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550095 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549984 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550027 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550018 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549964 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549979 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550074 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549927 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549955 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550093 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549963 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549960 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549962 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549910 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550073 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549941 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550103 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550083 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550014 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28549951 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550065 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28550040 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550053 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P09876531 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550068 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550089 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550035 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550079 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550088 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550052 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550031 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550029 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550078 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550027 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550091 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550045 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550093 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550047 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550058 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P09992245 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550028 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P09998755 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550069 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550062 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10040324 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550094 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550033 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550095 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550042 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10243527 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550037 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P09810601 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550049 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10111526 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550082 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550075 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550098 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P09967526 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550030 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550087 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550059 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550077 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10040322 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550097 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550083 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550084 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550102 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P09876537 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550100 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550071 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28550044 |
- |
- |