Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00473131_m1
          See other SIGLECE GE Assays ›
          Gene Symbol:
          Siglece
          Gene Name:
          sialic acid binding Ig-like lectin E
          Gene Aliases:
          Cd170, Siglec12, Siglec5, Siglec9, Siglecl1, mSiglec-E
          Chromosome Location:
          Chr.7: 43651070 - 43660179 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00473131_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Siglece
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031181.2 NP_112458.2 6 - 7 1228 59
          GenBank mRNA AF329269.1 - 6 - 7 1232 59
          AY348552.1 - 6 - 7 1237 59
          AY371487.1 - 6 - 7 1217 59
          BC023280.1 - 6 - 7 1227 3662856 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002951.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Siglece

          Panther Classification:

          Molecular Function -

          immunoglobulin superfamily cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          intracellular signal transduction
          negative regulation of inflammatory response
          negative regulation of phagocytosis, engulfment
          receptor activity
          receptor signaling protein activity
          carbohydrate binding
          sialic acid binding
          monosaccharide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132493 BC023280.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132493 NM_031181.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132493 AF329269.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 446758 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 357060 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60762 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 304142 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790418 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214562 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 655 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170904 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P132493 BC023280.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P132493 NM_031181.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P132493 AF329269.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P415766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25083514 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170904 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06178025 - -
          Whole Mouse Genome G4122A A_51_P415766 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125040 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132493 BC023280.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132493 NM_031181.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132493 AF329269.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170904 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05893761 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06306735 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06306738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25083433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06047522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06306736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06031458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06138842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06138840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05893753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06047519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06031457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06306738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06346129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06071552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06656153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06138841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25083502 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10562728 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562723 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562727 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562725 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562726 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562724 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562721 - -
          Affymetrix HT MG-430 PM Array Plate 1421463_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10562720 - -
          Mouse Genome 430 2.0 Array 1421463_at - -
          Mouse Genome 430 2.0 Array 1421463_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83382_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83382_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700026152.mm.1 TC0700002951.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026152.mm.1 NM_031181.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026152.mm.1 NM_031181.2 Refseq
          Mouse Expression Set 430 1421463_at - -
          Mouse Expression Set 430 1421463_PM_at - -
          Mouse Genome 430A 2.0 Array 1421463_PM_at - -
          Mouse Genome 430A 2.0 Array 1421463_at - -
          Affymetrix HT Mouse Genome 430A Array 1421463_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421463_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 434546 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 770524 - -
          Illumina mouse-6 v1.0 expression beadchip 770524 - -
          Illumina mouse-6 v1.0 expression beadchip 5550685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000770524 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251152 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2599192 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2599192 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251152 - -
          mouse-6 v1.1 (Illumina) scl000105.1_286-S - -
          mouse-6 v1.1 (Illumina) scl00095.1_110-S - -
          Illumina Sentrix mouse-6.v1 bead array 5550685 - -
          Illumina Sentrix mouse-6.v1 bead array 770524 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00095.1_110-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000105.1_286-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 770524 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550685 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1251152 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2599192 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2969577 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2599192 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251152 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2969577 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline