Search
Search

Chrdl1| Gene Symbol: Chrdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114385.1 | NP_001107857.1 | 4 - 5 | 814 | 76 | |
| NM_031258.3 | NP_112548.2 | 4 - 5 | 814 | 76 | ||
| GenBank mRNA | AF296451.1 | - | 4 - 5 | 736 | 76 | |
| AF305714.1 | - | 3 - 4 | 353 | 76 | ||
| AF321853.1 | - | 3 - 4 | 290 | 76 | ||
| AK142805.1 | - | 4 - 5 | 438 | 76 | ||
| AK164661.1 | - | 4 - 5 | 409 | 76 | ||
| BC050818.1 | - | 3 - 4 | 304 | 76 | ||
| BC066832.1 | - | 4 - 5 | 448 | 30102312 | 76 | |
Set Membership: |
|