Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00473300_m1
          See other SYTL1 GE Assays ›
          Gene Symbol:
          Sytl1
          Gene Name:
          synaptotagmin-like 1
          Gene Aliases:
          9030417L18, PSGL-1, Slp1
          Chromosome Location:
          Chr.4: 133253090 - 133263087 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00473300_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sytl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031393.2 NP_113570.2 2 - 3 331 83
          GenBank mRNA AB050741.1 - 2 - 3 310 83
          AK078868.1 - 2 - 3 319 83
          AK131162.1 - 2 - 3 288 83
          AK154176.1 - 2 - 3 326 83
          BC005623.1 - 2 - 3 331 3155535 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003694.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sytl1

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          exocytosis
          synaptic vesicle exocytosis
          vesicle-mediated transport
          regulation of calcium ion-dependent exocytosis
          calcium ion-regulated exocytosis of neurotransmitter
          calcium ion binding
          protein binding
          calcium-dependent phospholipid binding
          Rab GTPase binding
          syntaxin binding
          clathrin binding
          neurexin family protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163561 AK078868.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163561 AK131162.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163561 NM_031393.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2143693 AK078868.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2143693 AK131162.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2143693 NM_031393.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2143693 BC005623.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 610181 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40462 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 592227 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 267633 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 909812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 775338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13248 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163561 AK078868.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163561 AK131162.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163561 NM_031393.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2143693 AK078868.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2143693 AK131162.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2143693 NM_031393.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2143693 BC005623.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P485731 AK154176.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P485731 BC005623.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P485731 AB050741.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P485731 AK078868.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P485731 AK131162.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23316669 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00992835 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05408943 - -
          Whole Mouse Genome G4122A A_51_P485731 AK154176.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485731 BC005623.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485731 AB050741.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485731 AK078868.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485731 AK131162.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134502 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00992835 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03461834 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03879782 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03981736 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03879775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03879782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03879793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04001454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03981736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03267568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04081320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03981735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04001439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04001447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04046863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03879770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03879788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04001446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04081312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03879766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04001440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03879769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03451412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04081315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04001453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04081307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03451411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04001438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03451405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03879775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04001458 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10517019 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517025 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517022 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517021 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517011 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517020 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517017 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517010 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517016 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517014 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517027 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517013 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517023 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517024 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517012 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517018 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517026 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517015 - -
          Affymetrix HT MG-430 PM Array Plate 1450240_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10517009 - -
          Mouse Genome 430 2.0 Array 1450240_PM_a_at - -
          Mouse Genome 430 2.0 Array 1450240_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 269589_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 269589_at - -
          Affymetrix Mouse Clariom S Array PSR0400030282.mm.1 TC0400003694.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030282.mm.1 NM_031393.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030282.mm.1 NM_031393.2 Refseq
          Mouse Expression Set 430 1450240_PM_a_at - -
          Mouse Expression Set 430 1450240_a_at - -
          Murine Genome U74v2 Set 129830_g_at - -
          Murine Genome U74v2 Set 129830_PM_g_at - -
          Murine Genome U74v2 Set 129829_PM_at - -
          Murine Genome U74v2 Set 129829_at - -
          Mouse Genome 430A 2.0 Array 1450240_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1450240_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450240_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450240_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 837857 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 510487 - -
          Illumina mouse-6 v1.0 expression beadchip 510487 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000510487 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2651054 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2651054 - -
          mouse-6 v1.1 (Illumina) scl23718.13.1_59-S - -
          Illumina Sentrix mouse-6.v1 bead array 510487 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23718.13.1_59-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 510487 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2651054 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2651054 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl23718.13.1_59-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0