Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00473485_m1
          See other MMP1A GE Assays ›
          Gene Symbol:
          Mmp1a
          Gene Name:
          matrix metallopeptidase 1a (interstitial collagenase)
          Gene Aliases:
          Mcol-A, Mcola
          Chromosome Location:
          Chr.9: 7464141 - 7476869 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00473485_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mmp1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032006.3 NP_114395.1 6 - 7 946 127
          GenBank mRNA AJ278462.1 - 6 - 7 900 127
          AK049552.1 - 6 - 7 947 127
          BC117756.2 - 6 - 7 946 40090601 127

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000048.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mmp1a

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          positive regulation of fibroblast migration
          positive regulation of smooth muscle cell migration
          collagen catabolic process
          positive regulation of protein oligomerization
          endopeptidase activity
          metalloendopeptidase activity
          calcium ion binding
          peptidase activity
          metallopeptidase activity
          zinc ion binding
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984546 AK049552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984546 BQ558168.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984546 NM_032006.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 473591 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 332697 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 31870 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600116 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 111587 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900034 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 28811 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147206 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143533 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984546 AK049552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984546 BQ558168.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984546 NM_032006.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185688 AK049552.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185688 BC117756.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185688 BQ558168.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185688 NM_032006.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26288091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26288093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26288062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26288094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26288081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26288068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26288063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26288090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26288076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26288067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26288085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26288061 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06693095 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143533 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147206 - -
          Whole Mouse Genome G4122A A_52_P549223 AK049552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P549223 BC117756.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P549223 NM_032006.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P549223 AJ278462.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185688 AK049552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185688 BC117756.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185688 BQ558168.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185688 NM_032006.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117354 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127261 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147206 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143533 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07809992 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07809989 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07826104 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07809993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26288078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07809989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07865335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07891036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08115457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07472044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26288081 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10583087 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583081 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583083 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583086 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583084 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583088 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583082 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583089 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583085 - -
          Affymetrix HT MG-430 PM Array Plate 1422175_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10583080 - -
          Mouse Genome 430 2.0 Array 1422175_at - -
          Mouse Genome 430 2.0 Array 1422175_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83995_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83995_at - -
          Affymetrix Mouse Clariom S Array PSR0900000381.mm.1 NM_032006.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000381.mm.1 NM_032006.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000381.mm.1 TC0900000048.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900000383.mm.1 NM_032006.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000383.mm.1 NM_032006.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000383.mm.1 TC0900000048.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900000378.mm.1 NM_032006.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000378.mm.1 NM_032006.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000378.mm.1 TC0900000048.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900000380.mm.1 NM_032006.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000380.mm.1 NM_032006.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000380.mm.1 TC0900000048.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900000379.mm.1 NM_032006.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000379.mm.1 NM_032006.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000379.mm.1 TC0900000048.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900000382.mm.1 NM_032006.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000382.mm.1 NM_032006.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000382.mm.1 TC0900000048.mm.1 MTA TC
          Mouse Expression Set 430 1422175_at - -
          Mouse Expression Set 430 1422175_PM_at - -
          Mouse Genome 430A 2.0 Array 1422175_PM_at - -
          Mouse Genome 430A 2.0 Array 1422175_at - -
          Affymetrix HT Mouse Genome 430A Array 1422175_at - -
          Affymetrix HT Mouse Genome 430A Array 1422175_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 525670 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5910288 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005910288 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226914 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226914 - -
          mouse-6 v1.1 (Illumina) scl083995.6_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 5910288 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl083995.6_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5910288 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3077034 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3156019 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226914 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3156019 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3077034 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-rsn79:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0