| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100023318.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023318.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023322.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023322.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023327.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023327.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023302.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023302.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023314.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023314.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023317.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023317.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023316.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023316.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023293.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023293.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023295.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023295.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023301.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023301.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023303.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023303.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023305.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023305.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023310.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023310.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023315.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023315.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023319.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023319.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023324.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023324.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023329.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023329.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023296.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023296.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023309.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023309.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023311.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023311.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023320.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023320.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023323.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023323.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023328.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023328.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023308.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023308.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023325.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023325.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023304.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023304.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023312.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023312.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023313.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023313.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023321.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023321.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023326.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023326.mm.1 |
TC1100002497.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023330.mm.1 |
NM_033374.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023330.mm.1 |
TC1100002497.mm.1 |
MTA TC |