Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00473805_m1
          See other UBQLN4 GE Assays ›
          Gene Symbol:
          Ubqln4
          Gene Name:
          ubiquilin 4
          Gene Aliases:
          A1Up, A1u, AI663987, CIP75, UBIN
          Chromosome Location:
          Chr.3: 88553677 - 88569725 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00473805_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ubqln4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033526.2 NP_277068.1 10 - 11 1724 80
          GenBank mRNA AB040050.1 - 10 - 11 1724 80
          AK150211.1 - 10 - 11 1661 80
          AK156515.1 - 10 - 11 1678 80
          AK216845.1 - 1 - 2 68 80
          BC017686.1 - 10 - 11 1686 4236887 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000730.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ubqln4

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of proteasomal ubiquitin-dependent protein catabolic process
          negative regulation of autophagosome maturation
          polyubiquitin binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22314484 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22314483 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P362161 AB040050.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P362161 AB040050.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 362588 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 951298 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 718443 AK145655.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342904 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 710696 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 417539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395018 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972337 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 178857 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 47205 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154418 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141396 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P362161 AB040050.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P362161 AB040050.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P362161 AB040050.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P362161 AB040050.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22314502 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141396 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04981225 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154418 - -
          Whole Mouse Genome G4122A A_51_P362161 AB040050.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P362161 AB040050.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107879 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141396 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154418 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02805589 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22314484 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22314483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22314581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02716832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02805589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02716836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02481767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02921312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02921314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02626715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02962074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02716830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02438290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02716831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02862078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02716835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02570773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02438285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02716833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03117025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03114833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02921329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02921325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02921313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02921311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02441451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02438288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02438289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22314521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03117023 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10493257 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493256 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493250 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493252 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493255 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493249 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493251 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493253 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493258 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493248 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493254 - -
          Affymetrix HT MG-430 PM Array Plate 1448692_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1448691_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10493247 - -
          Mouse Genome 430 2.0 Array 1448691_PM_at - -
          Mouse Genome 430 2.0 Array 1448692_at - -
          Mouse Genome 430 2.0 Array 1448692_PM_at - -
          Mouse Genome 430 2.0 Array 1448691_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 94232_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 94232_at - -
          Affymetrix Mouse Clariom S Array PSR0300005169.mm.1 NM_033526.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005169.mm.1 NM_033526.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005169.mm.1 TC0300000730.mm.1 MTA TC
          Mouse Expression Set 430 1448692_at - -
          Mouse Expression Set 430 1448691_PM_at - -
          Mouse Expression Set 430 1448692_PM_at - -
          Mouse Expression Set 430 1448691_at - -
          Murine Genome U74v2 Set 170926_PM_at - -
          Murine Genome U74v2 Set 170926_at - -
          Mouse Genome 430A 2.0 Array 1448692_at - -
          Mouse Genome 430A 2.0 Array 1448691_at - -
          Mouse Genome 430A 2.0 Array 1448691_PM_at - -
          Mouse Genome 430A 2.0 Array 1448692_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448691_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448692_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448692_at - -
          Affymetrix HT Mouse Genome 430A Array 1448691_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 598655 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2970278 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002970278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2605268 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2605268 - -
          mouse-6 v1.1 (Illumina) scl22997.10_102-S - -
          Illumina Sentrix mouse-6.v1 bead array 2970278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl22997.10_102-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2970278 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2605268 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2605268 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl22997.10_102-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline