Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00474141_m1
          See other GPRC5D GE Assays ›
          Gene Symbol:
          Gprc5d
          Gene Name:
          G protein-coupled receptor, family C, group 5, member D
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 135105991 - 135137443 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00474141_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gprc5d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053118.1 NP_444348.1 2 - 3 1042 85
          GenBank mRNA AB025222.1 - 1 - 2 888 85
          AB099816.1 - 1 - 2 933 85
          AF218809.1 - 2 - 3 1042 85
          BC119192.1 - 2 - 3 1002 8733941 85
          BC119194.1 - 2 - 3 1002 8733943 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003313.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gprc5d

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          keratinization
          hair cycle
          molecular_function
          signal transducer activity
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009001 AB025221.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009001 AB025222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009001 AB099816.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009001 AF218809.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009001 AK137739.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178698 AB025221.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178698 AB025222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178698 AB099816.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178698 AF218809.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178698 AK137739.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 446770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 514768 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397285 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 424449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 742062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 1215 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153848 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130208 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009001 AB025221.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009001 AB025222.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009001 AB099816.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009001 AF218809.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009001 AK137739.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178698 AB025221.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178698 AB025222.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178698 AB099816.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178698 AF218809.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178698 AK137739.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P433333 BC119194.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P433333 BC119192.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P433333 AB025221.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P433333 AB025222.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P433333 AB099816.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24865234 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130208 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153848 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06077626 - -
          Whole Mouse Genome G4122A A_51_P433333 BC119194.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433333 BC119192.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433333 AB025221.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433333 AB025222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433333 AB099816.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113796 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153848 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130208 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05105251 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05105256 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05105262 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05105250 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05105259 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05105247 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05105255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24865198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05318893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05105250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05212419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05191262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05105567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05795123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05105248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05318890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05105247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05058426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05458696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05318884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05212428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05693329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05550480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05318883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05458697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05318879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05105251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05105256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05212429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05212420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05191261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05318886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05318881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05212425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24865190 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548757 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548760 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548756 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548758 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548755 - -
          Affymetrix HT MG-430 PM Array Plate 1420538_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10548754 - -
          Mouse Genome 430 2.0 Array 1420538_at - -
          Mouse Genome 430 2.0 Array 1420538_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93746_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93746_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600025984.mm.1 TC0600003313.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025984.mm.1 NM_001205396.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025984.mm.1 NM_001205396.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025984.mm.1 NM_053118.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025984.mm.1 NM_053118.1 Refseq
          Mouse Expression Set 430 1420538_PM_at - -
          Mouse Expression Set 430 1420538_at - -
          Mouse Genome 430A 2.0 Array 1420538_at - -
          Mouse Genome 430A 2.0 Array 1420538_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420538_at - -
          Affymetrix HT Mouse Genome 430A Array 1420538_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 575091 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4760021 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004760021 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2594738 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2594738 - -
          mouse-6 v1.1 (Illumina) scl28294.4.1_121-S - -
          Illumina Sentrix mouse-6.v1 bead array 4760021 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl28294.4.1_121-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4760021 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline