Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00474458_s1
          See other PCDHB15 GE Assays ›
          Gene Symbol:
          Pcdhb15
          Gene Name:
          protocadherin beta 15
          Gene Aliases:
          Pcdhb7, PcdhbO
          Chromosome Location:
          Chr.18: 37473546 - 37476340 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00474458_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pcdhb15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053140.3 NP_444370.1 1 324 70
          GenBank mRNA AK038663.1 - 1 326 70
          AY013776.1 - 1 153 70
          BC066178.1 - 1 330 30100073 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000372.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pcdhb15

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Process(es)

          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159833 BC066178.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159833 AK038663.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159833 AY013776.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159833 NM_053140.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159833 BC066178.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159833 AK038663.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159833 AY013776.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159833 NM_053140.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387928 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 433551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 28901 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142081 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159833 BC066178.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159833 AK038663.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159833 AY013776.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159833 NM_053140.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159833 BC066178.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159833 AK038663.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159833 AY013776.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159833 NM_053140.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159835 BC066178.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159835 AK038663.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159835 AY013776.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159835 NM_053140.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159833 BC066178.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159833 AK038663.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159833 AY013776.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159833 NM_053140.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159833 BC066178.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159833 AK038663.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159833 AY013776.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159833 NM_053140.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628444 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142081 - -
          Whole Mouse Genome G4122A A_51_P159835 BC066178.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159835 AK038663.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159835 AY013776.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159835 NM_053140.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159833 BC066178.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159833 AK038663.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159833 AY013776.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159833 NM_053140.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159833 BC066178.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159833 AK038663.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159833 AY013776.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159833 NM_053140.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107677 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142081 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13746217 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13746200 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13746220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13746221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13746210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13746219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13746208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628458 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10455105 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455107 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455106 - -
          Affymetrix HT MG-430 PM Array Plate 1443421_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1421549_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10455104 - -
          Mouse Genome 430 2.0 Array 1443421_PM_s_at - -
          Mouse Genome 430 2.0 Array 1421549_PM_at - -
          Mouse Genome 430 2.0 Array 1443421_s_at - -
          Mouse Genome 430 2.0 Array 1421549_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93886_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93886_at - -
          Affymetrix Mouse Clariom S Array PSR1800002852.mm.1 NM_053140.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002852.mm.1 NM_053140.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002852.mm.1 TC1800000372.mm.1 MTA TC
          Mouse Expression Set 430 1421549_PM_at - -
          Mouse Expression Set 430 1421549_at - -
          Mouse Expression Set 430 1443421_s_at - -
          Mouse Expression Set 430 1443421_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1421549_PM_at - -
          Mouse Genome 430A 2.0 Array 1421549_at - -
          Affymetrix HT Mouse Genome 430A Array 1421549_at - -
          Affymetrix HT Mouse Genome 430A Array 1421549_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443421_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1443421_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 729136 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 520725 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000520725 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2600952 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2600952 - -
          mouse-6 v1.1 (Illumina) scl093886.1_50-S - -
          Illumina Sentrix mouse-6.v1 bead array 520725 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl093886.1_50-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 520725 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2600952 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2600952 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline