Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00474485_s1
          See other PCDHB16 GE Assays ›
          Gene Symbol:
          Pcdhb16
          Gene Name:
          protocadherin beta 16
          Gene Aliases:
          Pcdhb8, PcdhbP
          Chromosome Location:
          Chr.18: 37477768 - 37483038 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00474485_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pcdhb16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053141.3 NP_444371.3 1 677 102
          GenBank mRNA AK030612.1 - 1 632 102
          AK036829.1 - 1 608 102
          AY013777.1 - 1 456 102
          BC115720.1 - 1 581 40086527 102
          BC116625.1 - 1 581 40086525 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000373.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pcdhb16

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s)

          chemical synaptic transmission
          synapse assembly
          calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072522 AK036267.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072522 AK036829.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072522 BC115720.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072522 BC116625.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072522 NM_053141.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 321625 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 823435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155705 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105519 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072522 AK036267.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072522 AK036829.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072522 BC115720.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072522 BC116625.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072522 NM_053141.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P191743 AK036267.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P191743 AK036829.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P191743 AY013777.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P191743 BC115720.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P191743 BC116625.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P24851 AK036267.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P24851 AK036829.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P24851 BC115720.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P24851 BC116625.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P24851 NM_053141.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628480 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08968275 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105519 - -
          Whole Mouse Genome G4122A A_52_P24851 AK036267.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P24851 AK036829.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P24851 BC115720.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P24851 BC116625.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P24851 NM_053141.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P191743 AK036267.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P191743 AK036829.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P191743 AY013777.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P191743 BC115720.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P191743 BC116625.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127139 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105519 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14095537 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13712927 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13804329 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13712932 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13712954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14092751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14095537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13804329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13712952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628467 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10455110 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455109 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455111 - -
          Affymetrix HT MG-430 PM Array Plate 1450216_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441358_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441994_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10455108 - -
          Mouse Genome 430 2.0 Array 1441994_PM_at - -
          Mouse Genome 430 2.0 Array 1441358_PM_at - -
          Mouse Genome 430 2.0 Array 1441994_at - -
          Mouse Genome 430 2.0 Array 1441358_at - -
          Mouse Genome 430 2.0 Array 1450216_PM_at - -
          Mouse Genome 430 2.0 Array 1450216_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93887_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93887_at - -
          Affymetrix Mouse Clariom S Array PSR1800002854.mm.1 NM_053141.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002854.mm.1 NM_053141.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002854.mm.1 TC1800000373.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002855.mm.1 NM_053141.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002855.mm.1 NM_053141.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002855.mm.1 TC1800000373.mm.1 MTA TC
          Mouse Expression Set 430 1450216_PM_at - -
          Mouse Expression Set 430 1450216_at - -
          Mouse Expression Set 430 1441358_PM_at - -
          Mouse Expression Set 430 1441358_at - -
          Mouse Expression Set 430 1441994_PM_at - -
          Mouse Expression Set 430 1441994_at - -
          Mouse Genome 430A 2.0 Array 1450216_PM_at - -
          Mouse Genome 430A 2.0 Array 1450216_at - -
          Affymetrix HT Mouse Genome 430A Array 1450216_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450216_at - -
          Affymetrix HT Mouse Genome 430B Array 1441358_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441358_at - -
          Affymetrix HT Mouse Genome 430B Array 1441994_at - -
          Affymetrix HT Mouse Genome 430B Array 1441994_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 545367 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4200048 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004200048 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2658008 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2658008 - -
          mouse-6 v1.1 (Illumina) scl52057.1_223-S - -
          Illumina Sentrix mouse-6.v1 bead array 4200048 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52057.1_223-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4200048 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2658008 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2658008 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0