Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00474511_s1
          See other PCDHB17 GE Assays ›
          Gene Symbol:
          Pcdhb17
          Gene Name:
          protocadherin beta 17
          Gene Aliases:
          Pcdhb16, PcdhbQ, mKIAA1621
          Chromosome Location:
          Chr.18: 37485021 - 37488290 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00474511_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pcdhb17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053142.3 NP_444372.2 1 893 80
          GenBank mRNA AF326310.1 - 1 755 80
          AK077255.1 - 1 864 80
          AK122533.1 - 1119 80
          AY013778.1 - 1 755 80
          BC017149.1 - 1 903 4506788 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000374.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pcdhb17

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Process(es)

          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P337771 BC017149.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P337771 AF326310.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P337771 AK077255.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P337771 AK122533.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P337771 NM_053142.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P337771 BC017149.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P337771 AF326310.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P337771 AK077255.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P337771 AK122533.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P337771 NM_053142.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342831 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 175937 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107051 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P337771 BC017149.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P337771 AF326310.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P337771 AK077255.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P337771 AK122533.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P337771 NM_053142.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P337771 BC017149.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P337771 AF326310.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P337771 AK077255.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P337771 AK122533.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P337771 NM_053142.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P337771 BC017149.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P337771 AF326310.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P337771 AK077255.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P337771 AK122533.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P337771 NM_053142.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P337771 BC017149.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P337771 AF326310.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P337771 AK077255.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P337771 AK122533.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P337771 NM_053142.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P180741 BC017149.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P180741 AF326310.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P180741 AK077255.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P180741 AK122533.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P180741 NM_053142.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628551 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08968294 - -
          Whole Mouse Genome G4122A A_51_P337771 BC017149.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337771 AF326310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337771 AK077255.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337771 AK122533.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337771 NM_053142.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337771 BC017149.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337771 AF326310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337771 AK077255.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337771 AK122533.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P337771 NM_053142.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180741 BC017149.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180741 AF326310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180741 AK077255.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180741 AK122533.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180741 NM_053142.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117950 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14009124 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14009147 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14009083 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14009145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14009160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14009083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14009155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13821107 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10455116 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455115 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455113 - -
          Affymetrix HT MG-430 PM Array Plate 1448933_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10455112 - -
          Mouse Genome 430 2.0 Array 1448933_at - -
          Mouse Genome 430 2.0 Array 1448933_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93888_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93888_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800002859.mm.1 NM_053142.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002859.mm.1 NM_053142.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002859.mm.1 TC1800000374.mm.1 MTA TC
          Mouse Expression Set 430 1448933_at - -
          Mouse Expression Set 430 1448933_PM_at - -
          Murine Genome U74v2 Set 168300_PM_r_at - -
          Murine Genome U74v2 Set 168390_PM_f_at - -
          Murine Genome U74v2 Set 168390_f_at - -
          Murine Genome U74v2 Set 168300_r_at - -
          Mouse Genome 430A 2.0 Array 1448933_PM_at - -
          Mouse Genome 430A 2.0 Array 1448933_at - -
          Affymetrix HT Mouse Genome 430A Array 1448933_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448933_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 752340 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2100725 - -
          Illumina mouse-6 v1.0 expression beadchip 2100725 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002100725 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1214296 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214296 - -
          mouse-6 v1.1 (Illumina) scl093888.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 2100725 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl093888.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2100725 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1214296 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1214296 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0