Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00475131_m1
          See other PRSS28 GE Assays ›
          Gene Symbol:
          Prss28
          Gene Name:
          protease, serine 28
          Gene Aliases:
          Isp1, mIsp-1
          Chromosome Location:
          Chr.17: 25308646 - 25311876 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00475131_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prss28
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053259.2 NP_444489.2 4 - 5 672 102
          GenBank mRNA AF184895.1 - 3 - 4 511 102
          AK144498.1 - 4 - 5 673 102
          BC119386.1 - 3 - 4 539 8734135 102
          BC119388.1 - 3 - 4 539 8734137 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000372.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prss28

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          embryo implantation
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023118 AK144498.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023118 NM_053259.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541926 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 443993 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165549 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023118 AK144498.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023118 NM_053259.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379837 AK144498.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379837 BC119386.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379837 BC119388.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379837 NM_053259.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P379837 AF184895.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102917 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165549 - -
          Whole Mouse Genome G4122A A_51_P379837 AK144498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379837 BC119386.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379837 BC119388.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379837 NM_053259.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P379837 AF184895.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113853 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165549 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13074895 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13074882 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13074885 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13074896 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13074903 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13074886 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13074900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13074885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13074886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13080951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13171540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13074881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13080956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13565252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13074903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13074895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13488471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13151992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13586965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13058810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13058816 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442778 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442772 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442775 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442774 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442777 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442773 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442776 - -
          Affymetrix HT MG-430 PM Array Plate 1450802_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442771 - -
          Mouse Genome 430 2.0 Array 1450802_at - -
          Mouse Genome 430 2.0 Array 1450802_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 114661_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 114661_at - -
          Affymetrix Mouse Clariom S Array PSR1700003076.mm.1 NM_053259.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003076.mm.1 NM_053259.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003076.mm.1 TC1700000372.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003074.mm.1 NM_053259.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003074.mm.1 NM_053259.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003074.mm.1 TC1700000372.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003075.mm.1 NM_053259.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003075.mm.1 NM_053259.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003075.mm.1 TC1700000372.mm.1 MTA TC
          Mouse Expression Set 430 1450802_PM_at - -
          Mouse Expression Set 430 1450802_at - -
          Mouse Genome 430A 2.0 Array 1450802_at - -
          Mouse Genome 430A 2.0 Array 1450802_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450802_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450802_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 651926 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4670427 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004670427 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2676226 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2676226 - -
          mouse-6 v1.1 (Illumina) scl50985.6.1_72-S - -
          Illumina Sentrix mouse-6.v1 bead array 4670427 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50985.6.1_72-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4670427 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2676226 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2676226 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline