Search
Search
| Gene Symbol: Vsx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054068.2 | NP_473409.1 | 2 - 3 | 702 | 68 | |
| GenBank mRNA | AF391758.1 | - | 2 - 3 | 547 | 68 | |
| AF395732.1 | - | 2 - 3 | 654 | 68 | ||
| AK044393.1 | - | 2 - 3 | 584 | 68 | ||
| AK044687.1 | - | 2 - 3 | 584 | 68 | ||
| AY056825.1 | - | 2 - 3 | 567 | 68 | ||
| BC057647.1 | - | 2 - 3 | 702 | 4511665 | 68 | |
| BC138456.1 | - | 2 - 3 | 632 | 8861476 | 68 | |
| BC138460.1 | - | 2 - 3 | 632 | 8861480 | 68 | |
Set Membership: |
|