Search
Search

Afg3l1| Gene Symbol: Afg3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054070.3 | NP_473411.2 | 14 - 15 | 1803 | 64 | |
| GenBank mRNA | AF329695.1 | - | 14 - 15 | 1744 | 64 | |
| AK159647.1 | - | 14 - 15 | 1767 | 64 | ||
| AK167964.1 | - | 14 - 15 | 1778 | 64 | ||
| BC056978.1 | - | 14 - 15 | 1762 | 6821746 | 64 | |
Set Membership: |
|