Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00475408_m1
          See other 1110038F14RIK GE Assays ›
          Gene Symbol:
          1110038F14Rik
          Gene Name:
          RIKEN cDNA 1110038F14 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 76948266 - 76950734 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00475408_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1110038F14Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_054099.2 NP_473440.1 3 - 4 607 87
          GenBank mRNA AK004154.1 - 3 - 4 561 87
          AK141496.1 - 3 - 4 399 87
          AK153059.1 - 4 - 5 561 87
          AK160695.1 - 3 - 4 630 87
          BC038324.1 - 3 - 4 607 1511597 87
          BC091734.1 - 3 - 4 372 3663572 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000615.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1110038F14Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351332 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351327 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351331 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351330 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351329 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351328 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351333 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P121447 AK141496.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P121447 AK153059.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P121447 AK004154.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P121447 AK160695.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P121447 BC038324.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P121447 AK141496.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P121447 AK153059.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P121447 AK004154.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P121447 AK160695.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P121447 BC038324.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355773 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114153 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113231 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P121447 AK141496.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P121447 AK153059.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P121447 AK004154.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P121447 AK160695.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P121447 BC038324.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P121447 AK141496.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P121447 AK153059.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P121447 AK004154.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P121447 AK160695.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P121447 BC038324.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351337 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113231 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114153 - -
          Whole Mouse Genome G4122A A_51_P121447 AK141496.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121447 AK153059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121447 AK004154.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121447 AK160695.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121447 BC038324.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121447 AK141496.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121447 AK153059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121447 AK004154.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121447 AK160695.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121447 BC038324.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135625 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137525 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113231 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114153 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12262050 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12262046 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12262041 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351328 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351330 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351332 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351331 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351327 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351329 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12049614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12063508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12456053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11859976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11945368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12225425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12209447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12209446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11856060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12419379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12262046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12262051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351297 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10425028 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425030 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425025 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425029 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425027 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425026 - -
          Affymetrix HT MG-430 PM Array Plate 1416885_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10425024 - -
          Mouse Genome 430 2.0 Array 1416885_PM_at - -
          Mouse Genome 430 2.0 Array 1416885_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 117171_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 117171_at - -
          Affymetrix Mouse Clariom S Array PSR1500004524.mm.1 NM_054099.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004524.mm.1 NM_054099.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004524.mm.1 TC1500000615.mm.1 MTA TC
          Mouse Expression Set 430 1416885_at - -
          Mouse Expression Set 430 1416885_PM_at - -
          Murine Genome U74v2 Set 94233_PM_at - -
          Murine Genome U74v2 Set 94233_at - -
          Mouse Genome 430A 2.0 Array 1416885_at - -
          Mouse Genome 430A 2.0 Array 1416885_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416885_at - -
          Affymetrix HT Mouse Genome 430A Array 1416885_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 380377 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1190075 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001190075 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2629375 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2629375 - -
          mouse-6 v1.1 (Illumina) scl47779.3.223_26-S - -
          Illumina Sentrix mouse-6.v1 bead array 1190075 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl47779.3.223_26-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1190075 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2863437 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2629375 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2863437 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2629375 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline