Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00475415_s1
          See other 2310034C09RIK GE Assays ›
          Gene Symbol:
          2310034C09Rik
          Gene Name:
          RIKEN cDNA 2310034C09 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 88758847 - 88759789 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00475415_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2310034C09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_054100.2 NP_473441.1 1 678 138
          GenBank mRNA AK009607.1 - 1 679 138
          BC064816.1 - 1 660 30287188 138

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000964.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 2310034C09Rik

          Panther Classification:

          Molecular Function -

          structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P488538 AK009607.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P488538 BC064816.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P488538 NM_054100.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P488538 AK009607.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P488538 BC064816.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P488538 NM_054100.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110593 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P488538 AK009607.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P488538 BC064816.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P488538 NM_054100.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P488538 AK009607.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P488538 BC064816.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P488538 NM_054100.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947826 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110593 - -
          Whole Mouse Genome G4122A A_51_P488538 AK009607.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488538 BC064816.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488538 NM_054100.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488538 AK009607.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488538 BC064816.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488538 NM_054100.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124218 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12569347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12982149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12869487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12569365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12869490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12493156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947844 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10436749 - -
          Affymetrix HT MG-430 PM Array Plate 1449986_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10436748 - -
          Mouse Genome 430 2.0 Array 1449986_PM_at - -
          Mouse Genome 430 2.0 Array 1449986_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 117172_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 117172_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600007563.mm.1 NM_054100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007563.mm.1 NM_054100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007563.mm.1 TC1600000964.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600007562.mm.1 NM_054100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007562.mm.1 NM_054100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007562.mm.1 TC1600000964.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600007559.mm.1 NM_054100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007559.mm.1 NM_054100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007559.mm.1 TC1600000964.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600007561.mm.1 NM_054100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007561.mm.1 NM_054100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007561.mm.1 TC1600000964.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600007560.mm.1 NM_054100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007560.mm.1 NM_054100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007560.mm.1 TC1600000964.mm.1 MTA TC
          Mouse Expression Set 430 1449986_PM_at - -
          Mouse Expression Set 430 1449986_at - -
          Mouse Genome 430A 2.0 Array 1449986_at - -
          Mouse Genome 430A 2.0 Array 1449986_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449986_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449986_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 817008 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4670750 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004670750 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228500 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228500 - -
          mouse-6 v1.1 (Illumina) scl48903.1.1_63-S - -
          Illumina Sentrix mouse-6.v1 bead array 4670750 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl48903.1.1_63-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4670750 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228500 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228500 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline