Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00475480_m1
          See other APOA5 GE Assays ›
          Gene Symbol:
          Apoa5
          Gene Name:
          apolipoprotein A-V
          Gene Aliases:
          1300007O05Rik, Apoav, RAP3
          Chromosome Location:
          Chr.9: 46268608 - 46271919 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00475480_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apoa5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080434.3 NP_536682.2 2 - 3 170 52
          GenBank mRNA AF327059.1 - 2 - 3 170 52
          AK004903.1 - 2 - 3 173 52
          AK004936.1 - 2 - 3 170 52
          AK050280.1 - 2 - 3 171 52
          BC011198.1 - 3 - 4 196 4196296 52
          BC029334.1 - 2 - 3 185 52

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000561.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apoa5

          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          triglyceride metabolic process
          cholesterol biosynthetic process
          transport
          lipid transport
          positive regulation of cholesterol esterification
          positive regulation of triglyceride catabolic process
          positive regulation of very-low-density lipoprotein particle remodeling
          triglyceride catabolic process
          regulation of intestinal cholesterol absorption
          neuron projection regeneration
          cholesterol efflux
          phospholipid efflux
          triglyceride-rich lipoprotein particle remodeling
          very-low-density lipoprotein particle remodeling
          high-density lipoprotein particle assembly
          lipoprotein metabolic process
          cholesterol homeostasis
          reverse cholesterol transport
          positive regulation of fatty acid biosynthetic process
          phosphatidylcholine metabolic process
          positive regulation of lipid catabolic process
          positive regulation of lipoprotein lipase activity
          acylglycerol homeostasis
          triglyceride homeostasis
          phospholipid binding
          enzyme activator activity
          heparin binding
          lipid binding
          cholesterol binding
          cholesterol transporter activity
          enzyme binding
          phosphatidylcholine binding
          lipase binding
          low-density lipoprotein particle receptor binding
          phosphatidylcholine-sterol O-acyltransferase activator activity
          lipase activator activity
          lipoprotein lipase activator activity
          lipoprotein particle receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26478880 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26478878 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26478879 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26478881 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049005 AK050280.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049005 NM_080434.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 10085 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 959804 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125743 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049005 AK050280.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049005 NM_080434.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P259930 BC029334.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P259930 AK004936.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P259930 AK050280.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P259930 NM_080434.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P259930 AF327059.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P292404 BC029334.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P292404 AK004903.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P292404 AK050280.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P292404 BC011198.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P292404 NM_080434.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478842 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125743 - -
          Whole Mouse Genome G4122A A_51_P259930 BC029334.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P259930 AK004936.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P259930 AK050280.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P259930 NM_080434.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P259930 AF327059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P292404 BC029334.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P292404 AK004903.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P292404 AK050280.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P292404 BC011198.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P292404 NM_080434.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133196 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125743 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07500999 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07500993 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07500978 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07500975 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07500984 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07500986 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07500987 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07500981 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26478881 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26478879 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26478878 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26478880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07662387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07500987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07979108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07592988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07500994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07501013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07500981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478874 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10585021 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585020 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585017 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585019 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585018 - -
          Affymetrix HT MG-430 PM Array Plate 1417610_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10585015 - -
          Mouse Genome 430 2.0 Array 1417610_PM_at - -
          Mouse Genome 430 2.0 Array 1417610_at - -
          Mouse Genome 430 2.0 Array 1419857_PM_at - -
          Mouse Genome 430 2.0 Array 1419857_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66113_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66113_at - -
          Affymetrix Mouse Clariom S Array PSR0900003940.mm.1 NM_080434.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003940.mm.1 TC0900000561.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900003939.mm.1 NM_080434.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003939.mm.1 TC0900000561.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900003938.mm.1 NM_080434.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003938.mm.1 TC0900000561.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900003935.mm.1 TC0900000561.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900003937.mm.1 NM_080434.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003937.mm.1 TC0900000561.mm.1 MTA TC
          Mouse Expression Set 430 1417610_at - -
          Mouse Expression Set 430 1417610_PM_at - -
          Murine Genome U74v2 Set 95727_PM_at - -
          Murine Genome U74v2 Set 95727_at - -
          Murine Genome U74v2 Set 95729_at - -
          Murine Genome U74v2 Set 95728_PM_g_at - -
          Murine Genome U74v2 Set 95728_g_at - -
          Murine Genome U74v2 Set 95729_PM_at - -
          Mouse Genome 430A 2.0 Array 1417610_PM_at - -
          Mouse Genome 430A 2.0 Array 1417610_at - -
          Affymetrix HT Mouse Genome 430A Array 1417610_at - -
          Affymetrix HT Mouse Genome 430A Array 1417610_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 802117 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6130471 - -
          Illumina mouse-6 v1.0 expression beadchip 6130471 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006130471 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2641301 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2641301 - -
          mouse-6 v1.1 (Illumina) scl37003.4_102-S - -
          Illumina Sentrix mouse-6.v1 bead array 6130471 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37003.4_102-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6130471 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2641301 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2641301 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline