Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00475529_m1
          See other MRPS2 GE Assays ›
          Gene Symbol:
          Mrps2
          Gene Name:
          mitochondrial ribosomal protein S2
          Gene Aliases:
          1500019M10Rik
          Chromosome Location:
          Chr.2: 28465956 - 28471177 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00475529_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrps2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001166031.2 NP_001159503.2 3 - 4 266 71
          NM_080452.3 NP_536700.3 3 - 4 323 71
          GenBank mRNA AB055389.1 - 3 - 4 314 71
          AK051184.1 - 3 - 4 323 71
          AK077219.1 - 4 - 5 492 71
          AK154131.1 - 3 - 4 317 71
          AK160129.1 - 3 - 4 315 71
          AK165956.1 - 3 - 4 314 71
          AK172120.1 - 2 - 3 786 71
          BC033460.1 - 2 - 3 772 71
          BC060503.1 - 3 - 4 299 6306321 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000482.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrps2

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          translation
          mitophagy in response to mitochondrial depolarization
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21091328 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21091331 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21091330 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198292 BC048377.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198292 BC060503.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198292 BC033460.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198292 AK171699.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198292 AK172120.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367565 BC048377.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367565 BC060503.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367565 AK206241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367565 BC033460.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367565 AK172120.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367565 BC048377.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367565 BC060503.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367565 AK206241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367565 BC033460.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367565 AK172120.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 771671 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670275 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269624 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121918 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367565 BC048377.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367565 BC060503.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367565 AK206241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367565 BC033460.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367565 AK172120.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367565 BC048377.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367565 BC060503.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367565 AK206241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367565 BC033460.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367565 AK172120.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198292 BC048377.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198292 BC060503.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198292 BC033460.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198292 AK171699.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198292 AK172120.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P181128 BC033460.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P181128 AK172120.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21091343 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121918 - -
          Whole Mouse Genome G4122A A_52_P181128 BC033460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P181128 AK172120.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P405955 BC048377.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P405955 BC060503.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P405955 AK184229.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P405955 BC033460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P405955 AK171699.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367565 BC048377.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367565 BC060503.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367565 AK206241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367565 BC033460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367565 AK172120.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367565 BC048377.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367565 BC060503.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367565 AK206241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367565 BC033460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367565 AK172120.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121918 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01204897 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21091328 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21091331 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21091330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01204897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01204890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01204912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01204911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01204908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01204889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01204899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01204895 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10470549 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470552 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470548 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470554 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470553 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470551 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470550 - -
          Affymetrix HT MG-430 PM Array Plate 1420846_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420845_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10470547 - -
          Mouse Genome 430 2.0 Array 1420846_PM_at - -
          Mouse Genome 430 2.0 Array 1420845_at - -
          Mouse Genome 430 2.0 Array 1420845_PM_at - -
          Mouse Genome 430 2.0 Array 1420846_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 118451_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 118451_at - -
          Affymetrix Mouse Clariom S Array PSR0200003378.mm.1 NM_001166031.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003378.mm.1 NM_001166031.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003378.mm.1 NM_080452.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003378.mm.1 NM_080452.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003378.mm.1 TC0200000482.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200003379.mm.1 NM_001166031.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003379.mm.1 NM_001166031.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003379.mm.1 NM_080452.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003379.mm.1 NM_080452.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003379.mm.1 TC0200000482.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200003380.mm.1 NM_001166031.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003380.mm.1 NM_001166031.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003380.mm.1 NM_080452.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003380.mm.1 NM_080452.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003380.mm.1 TC0200000482.mm.1 MTA TC
          Mouse Expression Set 430 1420846_at - -
          Mouse Expression Set 430 1420846_PM_at - -
          Mouse Expression Set 430 1420845_PM_at - -
          Mouse Expression Set 430 1420845_at - -
          Murine Genome U74v2 Set 160423_at - -
          Murine Genome U74v2 Set 160423_PM_at - -
          Mouse Genome 430A 2.0 Array 1420845_PM_at - -
          Mouse Genome 430A 2.0 Array 1420846_PM_at - -
          Mouse Genome 430A 2.0 Array 1420845_at - -
          Mouse Genome 430A 2.0 Array 1420846_at - -
          Affymetrix HT Mouse Genome 430A Array 1420845_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420846_at - -
          Affymetrix HT Mouse Genome 430A Array 1420846_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420845_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5860725 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005860725 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233225 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233225 - -
          mouse-6 v1.1 (Illumina) scl21129.4_153-S - -
          Illumina Sentrix mouse-6.v1 bead array 5860725 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21129.4_153-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5860725 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1233225 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233225 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline