Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00475567_m1
          See other POFUT1 GE Assays ›
          Gene Symbol:
          Pofut1
          Gene Name:
          protein O-fucosyltransferase 1
          Gene Aliases:
          O-FucT-1, mKIAA0180
          Chromosome Location:
          Chr.2: 153241532 - 153271029 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00475567_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pofut1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080463.3 NP_536711.3 1 - 2 193 64
          GenBank mRNA AF375885.1 - 1 - 2 162 64
          AK037857.1 - 1 - 2 157 64
          AK044629.1 - 1 - 2 171 64
          AK081059.1 - 1 - 2 150 64
          AK132301.1 - 1 - 2 148 64
          AK135635.1 - 1 - 2 175 64
          AK170038.1 - 1 - 2 148 64
          BC046295.1 - 1 - 2 193 6485896 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002312.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pofut1

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          somitogenesis
          carbohydrate metabolic process
          fucose metabolic process
          protein O-linked glycosylation
          Notch signaling pathway
          nervous system development
          heart development
          fucosylation
          protein O-linked fucosylation
          fucosyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups
          peptide-O-fucosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21753791 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21753793 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21753790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 868714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215826 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732499 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 828412 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 612722 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599945 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 372954 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 591414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 843529 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 68910 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 106115 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 668901 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 280133 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115740 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55816 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 69710 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 106864 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13333 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176411 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118409 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996299 CB521087.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996299 AK132301.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996299 NM_080463.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154570 BC046295.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154570 AK081059.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154570 AK132301.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154570 AK135635.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154570 AK170038.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21753808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21753811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21753794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21753805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21753807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21753796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21753800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21753799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21753806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21753790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21753814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21753815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21753816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21753813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21753791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21753802 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00529952 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00529942 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176411 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118409 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00529937 - -
          Whole Mouse Genome G4122A A_52_P183432 AK081059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P183432 AK132301.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P183432 NM_080463.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665833 AK081059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665833 AK132301.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665833 AK220326.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665833 AK044629.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665833 AK053648.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111705 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112108 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176411 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118409 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01645620 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01563207 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21753790 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21753793 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21753791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21753917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21753805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01162395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02045754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02045758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01563207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21753800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21753811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21753814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21753806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21753795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21753804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21753816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01645620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21753796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21753799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01273241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01676772 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10477289 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477293 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477290 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477294 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477296 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477288 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477292 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477295 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477287 - -
          Affymetrix HT MG-430 PM Array Plate 1450137_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1443881_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456323_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10477286 - -
          Mouse Genome 430 2.0 Array 1450137_at - -
          Mouse Genome 430 2.0 Array 1456323_at - -
          Mouse Genome 430 2.0 Array 1450137_PM_at - -
          Mouse Genome 430 2.0 Array 1456323_PM_at - -
          Mouse Genome 430 2.0 Array 1443881_at - -
          Mouse Genome 430 2.0 Array 1443881_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 140484_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 140484_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200018164.mm.1 NM_080463.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018164.mm.1 NM_080463.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018164.mm.1 TC0200002312.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200018165.mm.1 NM_080463.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018165.mm.1 NM_080463.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018165.mm.1 TC0200002312.mm.1 MTA TC
          Mouse Expression Set 430 1456323_at - -
          Mouse Expression Set 430 1456323_PM_at - -
          Mouse Expression Set 430 1450137_at - -
          Mouse Expression Set 430 1450137_PM_at - -
          Mouse Expression Set 430 1443881_at - -
          Mouse Expression Set 430 1443881_PM_at - -
          Murine Genome U74v2 Set 133811_PM_at - -
          Murine Genome U74v2 Set 133811_at - -
          Mouse Genome 430A 2.0 Array 1456323_at - -
          Mouse Genome 430A 2.0 Array 1450137_at - -
          Mouse Genome 430A 2.0 Array 1456323_PM_at - -
          Mouse Genome 430A 2.0 Array 1450137_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450137_at - -
          Affymetrix HT Mouse Genome 430A Array 1456323_at - -
          Affymetrix HT Mouse Genome 430A Array 1456323_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450137_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443881_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443881_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 638544 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1570458 - -
          Illumina mouse-6 v1.0 expression beadchip 1570458 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001570458 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2599360 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2599360 - -
          mouse-6 v1.1 (Illumina) scl00140484.1_147-S - -
          Illumina Sentrix mouse-6.v1 bead array 1570458 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00140484.1_147-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1570458 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2599360 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3132204 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3055425 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3132204 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3055425 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2599360 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline