Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00475794_m1
          See other PLIN2 GE Assays ›
          Gene Symbol:
          Plin2
          Gene Name:
          perilipin 2
          Gene Aliases:
          AA407157, ADPH, Adfp, Adrp
          Chromosome Location:
          Chr.4: 86626910 - 86670059 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00475794_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plin2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007408.3 NP_031434.3 4 - 5 446 79
          GenBank mRNA AK002911.1 - 4 - 5 382 79
          AK011014.1 - 4 - 5 435 79
          AK078967.1 - 3 - 4 422 79
          AK090277.1 - 4 - 5 377 79
          AK150177.1 - 4 - 5 383 79
          AK152081.1 - 4 - 5 382 79
          AK152263.1 - 4 - 5 374 79
          AK152877.1 - 4 - 5 382 79
          AK152896.1 - 4 - 5 374 79
          AK152982.1 - 4 - 5 406 79
          AK153222.1 - 4 - 5 404 79
          AK154582.1 - 4 - 5 383 79
          AK167140.1 - 4 - 5 384 79
          AY035850.1 - 3 - 4 305 79
          AY035851.1 - 3 - 4 305 79
          BC047124.1 - 4 - 5 370 79
          BC054766.1 - 4 - 5 359 6414796 79
          M93275.1 - 4 - 5 383 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002980.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Mouse lipid regulated genes 1700 Inventoried Mouse_lipid_regulated_genes Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plin2

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          long-chain fatty acid transport
          lipid storage
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999648 BC047124.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999648 BC054766.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999648 M93275.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999648 AY035850.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999648 AY035851.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 109325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 538571 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427261 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621672 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302436 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 128081 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92186 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106059 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148048 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999648 BC047124.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999648 BC054766.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999648 M93275.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999648 AY035850.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999648 AY035851.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P258150 BC047124.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P258150 M93275.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P258150 AK181873.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P258150 AK188078.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23081003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23081013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23081014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23081005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23081002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23081015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23081006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23081018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23081010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23081011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23081008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23081007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23081017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23080987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23081000 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148048 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106059 - -
          Whole Mouse Genome G4122A A_51_P258150 BC047124.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258150 M93275.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258150 AK181873.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258150 AK188078.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148048 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106059 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03776838 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03776839 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03776844 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03813106 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03776841 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03813103 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03813104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23080988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23081033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03593862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03813109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23081016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23081030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03311840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23081018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23081013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23081008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03776839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03776841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03622444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03673690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23081009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23081017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23081000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03311844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03999672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23081031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03931640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03813108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23081015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23080970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23081004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23081014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03813107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03813104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03776838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23081012 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10514229 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514222 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514227 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514225 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514230 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514226 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514224 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514231 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514228 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514223 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514232 - -
          Affymetrix HT MG-430 PM Array Plate 1448318_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10514221 - -
          Mouse Genome 430 2.0 Array 1448318_at - -
          Mouse Genome 430 2.0 Array 1448318_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11520_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11520_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400024149.mm.1 TC0400002980.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400024149.mm.1 NM_007408.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024149.mm.1 NM_007408.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024150.mm.1 TC0400002980.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400024150.mm.1 NM_007408.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024150.mm.1 NM_007408.3 Refseq
          Mouse Expression Set 430 1448318_at - -
          Mouse Expression Set 430 1448318_PM_at - -
          Murine Genome U74v2 Set 98589_PM_at - -
          Murine Genome U74v2 Set 98589_at - -
          Murine Genome U74v2 Set 135341_PM_r_at - -
          Murine Genome U74v2 Set 135341_r_at - -
          Mouse Genome 430A 2.0 Array 1448318_PM_at - -
          Mouse Genome 430A 2.0 Array 1448318_at - -
          Affymetrix HT Mouse Genome 430A Array 1448318_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448318_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 768506 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1740102 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001740102 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222246 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222246 - -
          mouse-6 v1.1 (Illumina) scl24151.12.434_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 1740102 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24151.12.434_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1740102 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222246 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222246 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl24151.12.434_2-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline