Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Arg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007482.3 | NP_031508.1 | 1 - 2 | 156 | 65 | |
GenBank mRNA | AK149471.1 | - | 1 - 2 | 139 | 65 | |
BC013341.1 | - | 1 - 2 | 133 | 4163626 | 65 | |
BC050005.1 | - | 1 - 2 | 170 | 3990246 | 65 | |
CB947842.1 | - | 1 - 2 | 160 | 65 | ||
CT010173.1 | - | 1 - 2 | 54 | 65 | ||
U51805.1 | - | 1 - 2 | 195 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |