Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00476035_s1
          See other ATOH1 GE Assays ›
          Gene Symbol:
          Atoh1
          Gene Name:
          atonal homolog 1 (Drosophila)
          Gene Aliases:
          Hath1, MATH-1, Math1, bHLHa14
          Chromosome Location:
          Chr.6: 64729146 - 64731243 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00476035_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atoh1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007500.4 NP_031526.1 1 329 64
          GenBank mRNA AK082354.1 - 1 351 64
          BC010820.1 - 1 329 4218223 64
          BC051256.1 - 1 347 6530849 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000754.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atoh1

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          Notch signaling pathway
          multicellular organism development
          nervous system development
          axon guidance
          brain development
          cerebral cortex development
          cell differentiation
          neuron differentiation
          inner ear morphogenesis
          auditory receptor cell differentiation
          auditory receptor cell fate specification
          auditory receptor cell fate determination
          negative regulation of apoptotic process
          positive regulation of auditory receptor cell differentiation
          regulation of neuron differentiation
          positive regulation of neuron differentiation
          positive regulation of transcription from RNA polymerase II promoter
          inner ear development
          positive regulation of inner ear receptor cell differentiation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          protein binding
          chromatin DNA binding
          sequence-specific DNA binding
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122015 BC051256.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122015 NM_007500.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122015 AK082354.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122015 BC010820.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122015 BC051256.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122015 NM_007500.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122015 AK082354.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122015 BC010820.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P383704 BC051256.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P383704 NM_007500.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P383704 AK082354.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P383704 BC010820.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P383704 BC051256.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P383704 NM_007500.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P383704 AK082354.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P383704 BC010820.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122015 BC051256.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122015 NM_007500.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122015 AK082354.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122015 BC010820.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24509153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24509169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24509168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24509167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24509173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24509154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24509162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24509163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24509156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24509159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24509164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24509170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24509173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05802705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05544536 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10538752 - -
          Affymetrix HT MG-430 PM Array Plate 1449822_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10538751 - -
          Mouse Genome 430 2.0 Array 1449822_PM_at - -
          Mouse Genome 430 2.0 Array 1449822_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11921_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11921_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600005657.mm.1 TC0600000754.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600005657.mm.1 NM_007500.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600005657.mm.1 NM_007500.4 Refseq
          Mouse Expression Set 430 1449822_at - -
          Mouse Expression Set 430 1449822_PM_at - -
          Murine Genome U74v2 Set 102569_PM_at - -
          Murine Genome U74v2 Set 102569_at - -
          Murine Genome U74v2 Set 168404_PM_at - -
          Murine Genome U74v2 Set 168404_at - -
          Mouse Genome 430A 2.0 Array 1449822_at - -
          Mouse Genome 430A 2.0 Array 1449822_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449822_at - -
          Affymetrix HT Mouse Genome 430A Array 1449822_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 621377 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 460278 - -
          Illumina mouse-6 v1.0 expression beadchip 460278 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000460278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2767705 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2767705 - -
          mouse-6 v1.1 (Illumina) scl0011921.1_156-S - -
          Illumina Sentrix mouse-6.v1 bead array 460278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0011921.1_156-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460278 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2767705 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2767705 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline