Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00476075_m1
          See other BAAT GE Assays ›
          Gene Symbol:
          Baat
          Gene Name:
          bile acid-Coenzyme A: amino acid N-acyltransferase
          Gene Aliases:
          AI118337, AI158864, BAT
          Chromosome Location:
          Chr.4: 49489416 - 49507912 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00476075_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Baat
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007519.3 NP_031545.2 2 - 3 751 88
          GenBank mRNA AK017923.1 - 2 - 3 701 88
          BC012683.1 - 2 - 3 698 88
          CB947692.1 - 1 - 2 76 88
          U95215.1 - 2 - 3 656 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002636.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Baat

          Panther Classification:

          Molecular Function -

          esterase hydrolase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          bile acid conjugation
          glycine metabolic process
          lipid metabolic process
          fatty acid metabolic process
          acyl-CoA metabolic process
          bile acid biosynthetic process
          bile acid metabolic process
          taurine metabolic process
          receptor binding
          palmitoyl-CoA hydrolase activity
          N-acyltransferase activity
          transferase activity
          transferase activity, transferring acyl groups
          hydrolase activity
          thiolester hydrolase activity
          glycine N-choloyltransferase activity
          carboxylic ester hydrolase activity
          medium-chain acyl-CoA hydrolase activity
          long-chain acyl-CoA hydrolase activity
          very long chain acyl-CoA hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 U95215.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 AK017923.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 BC012683.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 NM_007519.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 AA255212.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 U95215.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 AK017923.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 BC012683.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 NM_007519.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P43150 AA255212.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716304 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 441368 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 580401 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 512202 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207612 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 103082 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647033 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 101287 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8049 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174035 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130351 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100866 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 U95215.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 AK017923.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 BC012683.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 NM_007519.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 AA255212.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 U95215.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 AK017923.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 BC012683.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 NM_007519.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P43150 AA255212.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P245123 U95215.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22905447 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05231798 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05231820 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130351 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100866 - -
          Whole Mouse Genome G4122A A_52_P43150 U95215.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P43150 AK017923.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P43150 BC012683.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P43150 NM_007519.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P43150 AA255212.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P43150 U95215.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P43150 AK017923.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P43150 BC012683.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P43150 NM_007519.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P43150 AA255212.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P245123 U95215.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112408 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100866 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130351 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05231798 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03323081 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04023394 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04023398 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04023397 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03323084 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04023399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22905363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04023399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03246363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03961281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03590552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03420644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03961280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03323083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03420639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03246355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04023784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03632649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03961289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03314084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03961283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03246348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22905398 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10512897 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512900 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512898 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512899 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512896 - -
          Affymetrix HT MG-430 PM Array Plate 1419002_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1419001_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10512895 - -
          Mouse Genome 430 2.0 Array 1419002_PM_s_at - -
          Mouse Genome 430 2.0 Array 1419001_PM_at - -
          Mouse Genome 430 2.0 Array 1419001_at - -
          Mouse Genome 430 2.0 Array 1419002_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12012_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12012_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400021470.mm.1 TC0400002636.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021470.mm.1 NM_007519.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021470.mm.1 NM_007519.3 Refseq
          Mouse Expression Set 430 1419002_PM_s_at - -
          Mouse Expression Set 430 1419001_at - -
          Mouse Expression Set 430 1419001_PM_at - -
          Mouse Expression Set 430 1419002_s_at - -
          Murine Genome U74v2 Set 104273_PM_at - -
          Murine Genome U74v2 Set 104273_at - -
          Mouse Genome 430A 2.0 Array 1419002_s_at - -
          Mouse Genome 430A 2.0 Array 1419001_PM_at - -
          Mouse Genome 430A 2.0 Array 1419001_at - -
          Mouse Genome 430A 2.0 Array 1419002_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1419002_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1419001_at - -
          Affymetrix HT Mouse Genome 430A Array 1419001_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419002_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 565059 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 730739 - -
          Illumina mouse-6 v1.0 expression beadchip 730739 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000730739 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259009 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259009 - -
          mouse-6 v1.1 (Illumina) scl24338.5.1_130-S - -
          Illumina Sentrix mouse-6.v1 bead array 730739 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24338.5.1_130-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 730739 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1259009 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1259009 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0