Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30128863 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30128862 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P59681 |
NM_008287.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P59681 |
AK005016.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P59681 |
AW045776.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P59681 |
BC092375.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P59681 |
NM_008287.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P59681 |
AK005016.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P59681 |
AW045776.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P59681 |
BC092375.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
29241 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
284037 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
414906 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
541970 |
AB009377.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
541970 |
AK036094.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
541970 |
AK077619.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
541970 |
AK089267.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
541970 |
BC053052.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
541970 |
NM_007859.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305964 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
751487 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
30399 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
56325 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P130590 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P127810 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P59681 |
NM_008287.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P59681 |
AK005016.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P59681 |
AW045776.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P59681 |
BC092375.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P59681 |
NM_008287.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P59681 |
AK005016.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P59681 |
AW045776.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P59681 |
BC092375.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128845 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128843 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128833 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128837 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128848 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128853 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128856 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128841 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128852 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128862 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128832 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128835 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128855 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128839 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128851 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128859 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30128858 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08331597 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P130590 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P127810 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P59681 |
NM_008287.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P59681 |
AK005016.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P59681 |
AW045776.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P59681 |
BC092375.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P59681 |
NM_008287.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P59681 |
AK005016.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P59681 |
AW045776.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P59681 |
BC092375.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P102946 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P124419 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P127810 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P130590 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12282256 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12457997 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12458004 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30128862 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30128863 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128823 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128826 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128804 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128817 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128832 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128785 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128840 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128856 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128816 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128853 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128855 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128799 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128808 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128837 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128825 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128862 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128797 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128806 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128784 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128852 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128780 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128859 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128833 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128809 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128845 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128843 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128814 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30128830 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128856 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128821 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128840 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128779 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128843 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128855 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128777 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128848 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128824 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128853 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12213139 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128851 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128862 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128797 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128799 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128833 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128845 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128823 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128792 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128819 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128839 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128781 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128832 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128791 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12367771 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12456233 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128858 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12379601 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12213137 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128830 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128778 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12230774 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128837 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128822 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128826 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128802 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128841 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128795 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30128859 |
- |
- |