Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00476230_m1
          See other ARHGEF1 GE Assays ›
          Gene Symbol:
          Arhgef1
          Gene Name:
          Rho guanine nucleotide exchange factor (GEF) 1
          Gene Aliases:
          Lbcl2, Lsc
          Chromosome Location:
          Chr.7: 24902917 - 24926594 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00476230_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arhgef1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130150.1 NP_001123622.1 4 - 5 381 64
          NM_001130151.1 NP_001123623.1 4 - 5 381 64
          NM_001130152.1 NP_001123624.1 4 - 5 390 64
          NM_001130153.1 NP_001123625.1 4 - 5 381 64
          NM_008488.2 NP_032514.1 4 - 5 381 64
          GenBank mRNA AK012156.1 - 4 - 5 331 64
          AK043027.1 - 4 - 5 345 64
          AK051518.1 - 4 - 5 331 64
          AK131181.1 - 4 - 5 278 64
          BB840025.1 - 4 - 5 381 64
          BC012488.1 - 4 - 5 341 3154558 64
          CV783191.1 - 4 - 5 313 64
          U58203.1 - 4 - 5 311 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000403.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arhgef1

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of Rho protein signal transduction
          termination of G-protein coupled receptor signaling pathway
          negative regulation of cell death
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          GTPase activator activity
          protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700003021.mm.1 TC0700000403.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003021.mm.1 NM_001130150.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003021.mm.1 NM_001130150.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003021.mm.1 NM_001130151.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003021.mm.1 NM_001130151.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003021.mm.1 NM_001130152.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003021.mm.1 NM_001130152.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003021.mm.1 NM_001130153.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003021.mm.1 NM_001130153.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003021.mm.1 NM_008488.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003021.mm.1 NM_008488.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003017.mm.1 TC0700000403.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003017.mm.1 NM_001130150.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003017.mm.1 NM_001130150.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003017.mm.1 NM_001130151.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003017.mm.1 NM_001130151.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003017.mm.1 NM_001130152.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003017.mm.1 NM_001130152.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003017.mm.1 NM_001130153.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003017.mm.1 NM_001130153.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003017.mm.1 NM_008488.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003017.mm.1 NM_008488.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003020.mm.1 TC0700000403.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003020.mm.1 NM_001130150.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003020.mm.1 NM_001130150.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003020.mm.1 NM_001130151.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003020.mm.1 NM_001130151.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003020.mm.1 NM_001130152.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003020.mm.1 NM_001130152.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003020.mm.1 NM_001130153.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003020.mm.1 NM_001130153.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003020.mm.1 NM_008488.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003020.mm.1 NM_008488.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 652623 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline