Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00476781_s1
          See other PHXR4 GE Assays ›
          Gene Symbol:
          Phxr4
          Gene Name:
          per-hexamer repeat gene 4
          Gene Aliases:
          Sp5
          Chromosome Location:
          Chr.9: 13431361 - 13432751 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00476781_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Phxr4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028271.1 1 542 67
          GenBank mRNA X12806.1 - 1 554 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000086.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077088 X12806.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077088 NR_028271.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077088 BC107288.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077088 BC107289.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077088 BC127147.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 439836 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155078 - -
          Whole Mouse Genome G4122A A_51_P483215 X12806.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483215 NR_028271.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07842219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07504570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07862560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07959233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07644356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07842221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07745292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07612547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26311252 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10583204 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583206 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583205 - -
          Affymetrix HT MG-430 PM Array Plate 1422272_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10583203 - -
          Mouse Genome 430 2.0 Array 1422272_at - -
          Mouse Genome 430 2.0 Array 1422272_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900000614.mm.1 NR_028271.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000614.mm.1 NR_028271.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000614.mm.1 TC0900000086.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900000616.mm.1 NR_028271.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000616.mm.1 NR_028271.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000616.mm.1 TC0900000086.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900000615.mm.1 NR_028271.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000615.mm.1 NR_028271.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000615.mm.1 TC0900000086.mm.1 MTA TC
          Mouse Expression Set 430 1422272_PM_at - -
          Mouse Expression Set 430 1422272_at - -
          Murine Genome U74v2 Set 101700_PM_at - -
          Murine Genome U74v2 Set 101700_at - -
          Mouse Genome 430A 2.0 Array 1422272_PM_at - -
          Mouse Genome 430A 2.0 Array 1422272_at - -
          Affymetrix HT Mouse Genome 430A Array 1422272_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422272_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 470803 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1770161 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0