Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00477180_m1
          See other PSMC5 GE Assays ›
          Gene Symbol:
          Psmc5
          Gene Name:
          protease (prosome, macropain) 26S subunit, ATPase 5
          Gene Aliases:
          mSUG1
          Chromosome Location:
          Chr.11: 106256185 - 106263112 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00477180_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psmc5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008950.1 NP_032976.1 2 - 3 98 83
          GenBank mRNA AK010505.1 - 2 - 3 118 83
          AK012294.1 - 2 - 3 130 83
          AK135451.1 - 2 - 3 108 83
          AK145703.1 - 2 - 3 102 83
          BC030840.1 - 2 - 3 130 3994523 83
          Z54219.1 - 2 - 3 98 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001777.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psmc5

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          protein catabolic process
          ER-associated ubiquitin-dependent protein catabolic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          negative regulation of transcription, DNA-templated
          positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
          positive regulation of inclusion body assembly
          positive regulation of proteasomal protein catabolic process
          nucleotide binding
          receptor binding
          protein binding
          ATP binding
          transcription factor binding
          hydrolase activity
          ATPase activity
          TBP-class protein binding
          thyrotropin-releasing hormone receptor binding
          proteasome-activating ATPase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P657435 BC030840.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P657435 Z54219.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P657435 AK010505.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P657435 AK012294.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P657435 AK135451.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P657435 BC030840.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P657435 Z54219.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P657435 AK010505.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P657435 AK012294.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P657435 AK135451.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 455880 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358369 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 180280 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 419141 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117199 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P657435 BC030840.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P657435 Z54219.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P657435 AK010505.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P657435 AK012294.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P657435 AK135451.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P657435 BC030840.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P657435 Z54219.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P657435 AK010505.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P657435 AK012294.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P657435 AK135451.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P657435 BC030840.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P657435 Z54219.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P657435 AK010505.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P657435 AK012294.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P657435 AK135451.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P657435 BC030840.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P657435 Z54219.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P657435 AK010505.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P657435 AK012294.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P657435 AK135451.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232355 BC030840.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232355 Z54219.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232355 AK010505.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232355 AK012294.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232355 AK135451.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28100509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28100504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28100502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28100515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28100500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28100498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28100511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28100514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28100499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28100506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28100501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28100503 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07479257 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117199 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07479250 - -
          Whole Mouse Genome G4122A A_52_P657435 BC030840.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P657435 Z54219.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P657435 AK010505.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P657435 AK012294.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P657435 AK135451.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P657435 BC030840.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P657435 Z54219.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P657435 AK010505.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P657435 AK012294.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P657435 AK135451.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P232355 BC030840.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P232355 Z54219.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P232355 AK010505.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P232355 AK012294.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P232355 AK135451.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122061 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117199 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09133255 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09133253 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09133245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28100576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09699639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09088592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09133255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09088587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09186235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09088597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28100512 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10382072 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382079 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382075 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382070 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382071 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382073 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382078 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382080 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382077 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382074 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382076 - -
          Affymetrix HT MG-430 PM Array Plate 1415740_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10382069 - -
          Mouse Genome 430 2.0 Array 1415740_PM_at - -
          Mouse Genome 430 2.0 Array 1415740_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19184_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19184_at - -
          Affymetrix Mouse Clariom S Array PSR1100016805.mm.1 NM_008950.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016805.mm.1 NM_008950.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016805.mm.1 TC1100001777.mm.1 MTA TC
          Mouse Expression Set 430 1415740_PM_at - -
          Mouse Expression Set 430 1415740_at - -
          Mouse Expression Set 430 1415740_at - -
          Mouse Expression Set 430 1415740_PM_at - -
          Murine Genome U74v2 Set 160534_at - -
          Murine Genome U74v2 Set 160534_PM_at - -
          Murine Genome U74v2 Set 161921_PM_f_at - -
          Murine Genome U74v2 Set 161921_f_at - -
          Murine Genome U74v2 Set 171139_PM_at - -
          Murine Genome U74v2 Set 171139_at - -
          Mouse Genome 430A 2.0 Array 1415740_PM_at - -
          Mouse Genome 430A 2.0 Array 1415740_at - -
          Affymetrix HT Mouse Genome 430A Array 1415740_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1415740_at - -
          Affymetrix HT Mouse Genome 430B Array 1415740_at - -
          Affymetrix HT Mouse Genome 430B Array 1415740_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 834065 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6550021 - -
          Illumina mouse-6 v1.0 expression beadchip 2760315 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550021 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002760315 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220632 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251877 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220632 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251877 - -
          mouse-6 v1.1 (Illumina) scl40797.10.1_7-S - -
          mouse-6 v1.1 (Illumina) scl0001433.1_54-S - -
          Illumina Sentrix mouse-6.v1 bead array 2760315 - -
          Illumina Sentrix mouse-6.v1 bead array 6550021 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001433.1_54-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40797.10.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2760315 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550021 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1220632 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1251877 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251877 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220632 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40797.10.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0001433.1_54-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline